Skip to Content
Merck
All Photos(1)

Documents

MAK131

Sigma-Aldrich

Formaldehyde Assay

sufficient for 100 fluorometric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

usage

sufficient for 100 fluorometric tests

detection method

fluorometric

relevant disease(s)

cancer

storage temp.

2-8°C

General description

Formaldehyde is widely employed in industry for a wide range of applications. It acts as a disinfectant and biocide, and is utilized as a tissue fixative and embalming agent. Formaldehyde is a key metabolite produced during normal metabolism and contributes to methylation reactions and protein and nucleic acid synthesis. Increased production of formaldehyde has been observed in many cancer cell lines and increased formaldehyde concentration in urine has been associated with bladder and prostrate cancer.

Application


  • Modified Natural Rubber as a Simple Chemical Sensor with Smartphone Detection for Formaldehyde Content in a Seafood Sample.: This article explores the use of modified natural rubber as a chemical sensor, paired with smartphone detection, to measure formaldehyde content in seafood. This approach highlights the potential for accessible and mobile detection systems in food safety applications (Yeerum et al., 2022).

  • Optical enzymatic formaldehyde biosensor based on alcohol oxidase and pH-sensitive methacrylic-acrylic optode membrane.: The study develops an optical enzymatic biosensor for formaldehyde, using alcohol oxidase with a pH-sensitive membrane, offering a novel approach for quick and sensitive detection in various analytical applications (Nurlely et al., 2022).

Features and Benefits

Compatible with high-throughput handling systems. Can be adapted for use with cuvettes.

Suitability

Suitable for the detection of formaldehyde in urine and other biological samples.

Principle

The Formaldehyde Assay kit provides a simple and direct procedure for measuring formaldehyde levels in urine and other biological samples. In this assay, formaldehyde is derivatized with acetoacetanilide in the presence of ammonia resulting in a fluorescent (λex = 370/λem = 470 nm) product, proportional to the formaldehyde present.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A - STOT SE 1 - STOT SE 3

Target Organs

Eyes,Central nervous system, Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cristina A Nadalutti et al.
Scientific reports, 10(1), 5575-5575 (2020-03-30)
Formaldehyde (FA) is a simple biological aldehyde that is produced inside cells by several processes such as demethylation of DNA and proteins, amino acid metabolism, lipid peroxidation and one carbon metabolism (1-C). Although accumulation of excess FA in cells is
Lysine-specific demethylase 1 in breast cancer cells contributes to the production of endogenous formaldehyde in the metastatic bone cancer pain model of rats.
Liu J, et al.
PLoS ONE, 8(3), e58957-e58957 (2013)
Wancang Liu et al.
Frontiers in microbiology, 11, 1988-1988 (2020-09-26)
Traditional diosgenin manufacturing process has led to serious environmental contamination and wastewater. Clean processes are needed that can alternate the diosgenin production. The β-glucosidase FBG1, cloned from Fusarium sp. CPCC 400709, can biotransform trillin and produce diosgenin. In this study
Formaldehyde in the workplace and in the home: exploring its clinical toxicology.
Montanaro A.
Laboratory Medicine, 27(11), 752-758 (2015)
Wan-Cang Liu et al.
Microbial biotechnology, 12(6), 1226-1236 (2019-05-28)
Pichia pastoris KM71H (MutS ) is an efficient producer of hard-to-express proteins such as the membrane protein P-glycoprotein (Pgp), an ATP-powered efflux pump which is expressed properly, but at very low concentration, using the conventional induction strategy. Evaluation of different

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service