Skip to Content
Merck
All Photos(3)

Key Documents

MAK097

Sigma-Aldrich

Glucose Oxidase Activity Assay Kit

sufficient for 100 colorimetric or fluorometric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

usage

sufficient for 100 colorimetric or fluorometric tests

detection method

colorimetric
fluorometric

storage temp.

−20°C

General description

The new Glucose Oxidase Activity Assay Kit, MAK501, is now available! Glucose Oxidase (GOx, EC 1.1.3.4) is an enzyme found in insects, fungi, and bacteria that catalyzes the oxidation of D-glucose to D-gluconolactone. GOx is widely used in the food, beverage, chemical, and pharmaceutical industries.[1]

Suitability

Suitable for the measurement of glucose oxidase (GOx) in a variety of samples including cells and serum

Principle

The Glucose Oxidase Activity Assay Kit provides a simple and direct procedure for measuring GOx activity in a variety of biological samples. GOx activity is determined by a coupled enzyme assay, in which GOx oxidizes D-glucose resulting in the production of hydrogen peroxide (H2O2) that reacts with a probe, generating a colorimetric (570 nm)/fluorometric (λex = 535/λem = 587 nm) product, proportional to the GOx present. One unit of GOx is defined as the amount of enzyme that generates 1.0 μmole of H2O2 per minute at 37 °C.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3 - Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glucose oxidase ? An overview
Sandip B B, et al.
Biotechnology Advances, 27(4), 489-501 (2009)
DNA-guided assembly of a five-component enzyme cascade for enhanced conversion of cellulose to gluconic acid and H2O2.
Chen Q, et al.
Journal of Biotechnology, 263 , 30-35 (2017)
Glucagon-like peptide 1 receptor agonist ameliorates the insulin resistance function of islet β cells via the activation of PDX-1/JAK signaling transduction in C57/BL6 mice with high-fat diet-induced diabetes.
Hao T, et al.
International Journal of Molecular Medicine, 39(4), 1029-1036 (2017)
Umarah Zahoor Banday et al.
Ecotoxicology and environmental safety, 183, 109584-109584 (2019-08-26)
There is a dire need to assess the quality of fishes transported for human consumption as lately, their health is challenged because of anthropogenic activities. Heavy metals with a long environmental persistence are toxic to fishes and the humans. The
Suppression of a single BAHD gene in Setaria viridis causes large, stable decreases in cell wall feruloylation and increases biomass digestibility.
Souza W R, et al.
The New phytologist (2018)

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. Hello. May I know about this product's protocol? I'm interested to buy this but I have to know first about this protocol. Is it can be used for cell culture? If it's possible, can you send me this product's protocol through my email? Thank you.

    1 answer
    1. Glucose Oxidase Activity Assay Kit (Cat. no. MAK097) is suitable for the measurement of glucose oxidase in a variety of samples including cells and serum. Please see the Product Information Sheet available in the "Documentation" section of the product page: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/438/931/mak097bul.pdf.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service