Skip to Content
Merck
All Photos(2)

Documents

MAK020

Sigma-Aldrich

Fructose-6-Phosphate Assay Kit

sufficient for 100 fluorometric tests

Synonym(s):

Fructose-6-P Quantification Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

usage

sufficient for 100 fluorometric tests

detection method

fluorometric

relevant disease(s)

cancer

storage temp.

−20°C

General description

Fructose-6-phosphate (F6P) is a glycolytic pathway intermediate produced by the isomerization of glucose-6-phosphate by phosphoglucose isomerase. F6P is further phosphorylated to fructose 1,6-biphosphate which is subsequently cleaved to glyceraldehyde phosphate and dihydroxyacetone phosphate. F6P can also be shunted from glycolysis to the non-oxidative branch of the pentose phosphate pathway as a means of generative pentose phosphates for nucleotide synthesis. F6P levels are elevated in rapidly proliferating cells such as cancer cells. The Fructose-6-phosphate Assay Kit is a highly sensitive and simple fluorescence-based method of quantifying F6P in a variety of samples.

Application

Fructose-6-Phosphate Assay Kit has been used to measure fructose-6-phosphate (F-6-P) and glucose-6-phosphate (G6P).

Suitability

Suitable for quantifying Fructose-6-Phosphate in a variety of samples

Principle

Fructose-6-phosphate concentration is determined by a coupled enzyme reaction, which results in a fluorometric (λex = 535 nm/λem = 587 nm) product, proportional to the fructose-6-phosphate present. Typical sensitivities for this kit are 0.01 to 0.5 nmoles of F6P.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Minle Li et al.
Cell death & disease, 13(2), 99-99 (2022-02-04)
Transketolase (TKT) which is an important metabolic enzyme in the pentose phosphate pathway (PPP) participates in maintaining ribose 5-phosphate levels. TKT is necessary for maintaining cell growth. However, we found that in addition to this, TKT can also affect tumor
Reciprocal regulation between O-GlcNAcylation and tribbles pseudokinase 2 (TRIB2) maintains transformative phenotypes in liver cancer cells.
Yao B, et al.
Cellular Signalling, 28(11) (2016)
Kenichi Matsumoto et al.
Cancer letters, 500, 29-40 (2020-12-12)
Glycolysis emerges as a new therapeutic target for malignancies. The inhibition of glycolytic activator, PFKFB3, repairs tumor endothelial cell function, and normalizing the tumor microenvironment. We aimed to investigate the significance of PFKFB3 in HCC, and the effects of the
Proteomic and biochemical basis for enhanced growth yield of Enterobacter sp. LCR1 on insoluble phosphate medium.
Kumar A and Rai L C
Microbiological research, 170 (2015)
Sicheng Gao et al.
DNA and cell biology, 38(4), 286-296 (2019-02-15)
Abnormal expression of O-Linked β-N-acetylglucosamine (O-GlcNAc) and β-catenin is a general feature of cancer and contributes to transformed phenotypes. In this study, we identified the interaction between O-GlcNAc and β-catenin, and explored their effects on the progression of liver cancer.

Articles

Warburg effect enhances glucose to lactate conversion in tumor cells, regardless of oxygen levels; impacting cancer metabolism since 1924.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service