Skip to Content
Merck
All Photos(2)

Key Documents

MAK003

Sigma-Aldrich

Branched Chain Amino Acid Kit

sufficient for 100 colorimetric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

usage

sufficient for 100 colorimetric tests

detection method

colorimetric

relevant disease(s)

endocrinological disorders, diabetes; gastrointestinal diseases; neurological disorders

storage temp.

−20°C

General description

The branched-chain amino acids (BCAA), leucine, isoleucine, and valine, are the most common essential amino acids in proteins. The BCAA may function in both energy production and as nutrient signals.[1] Dietary supplementation with BCAA may aid in the treatment of various disorders such as Amyotrophic Lateral Sclerosis (ALS), latent and chronic hepatic encephalopathies,[2] and to prevent muscle atrophy in cancer patients. BCAA may increase the rate of protein synthesis and sensitize cells to insulin and insulin-like growth factor.[3]

Application

Branched Chain Amino Acid Kit has been used to determine the concentration of hepatic branched-chain amino acids, in a study assessing the role of CREB regulated transcription coactivator 2 (CRTC2) in hepatic lipid homeostasis and obesity in mice.[4]

Suitability

Suitable for measuring Branched Chain Amino Acids (BCAA′s) in a variety of biological samples.

Principle

In this assay, BCAA concentration is determined using a coupled enzyme reaction, which results in a colorimetric (450 nm) product, proportional to the BCAA present. The BCAA Assay Kit provides a simple convenient means of measuring the BCAAs in a variety of biological samples.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Protein substitution to produce a processed cheese with high branched-chain amino acids of medical and genetic importance.
El-Shazly H A M, et al.
The Egyptian journal of medical human genetics., 11(2), 121-133 (2010)
Hager M Kowash et al.
Clinical science (London, England : 1979), 136(15), 1117-1137 (2022-07-20)
Maternal infection during pregnancy increases the offspring risk of developing a variety of neurodevelopmental disorders (NDDs), including schizophrenia. While the mechanisms remain unclear, dysregulation of placental function is implicated. We hypothesised that maternal infection, leading to maternal immune activation and
Aidan M Sokolov et al.
Human molecular genetics, 29(18), 3003-3013 (2020-08-22)
Pathogenic mutations in the solute carrier family 7 member 5 (SLC7A5) gene, which encodes an amino acid transporter cause microcephaly and seizures, yet the mechanisms responsible for these phenotypes are unclear. Models have demonstrated that Slc7a5 deletion is embryonic lethal
Multifaceted role of insulin-like growth factors and mammalian target of rapamycin in skeletal muscle.
Frost R A and Charles H L
Endocrinol. Metab. Clin., 41(2), 297-322 (2012)
Itziar Lamiquiz-Moneo et al.
Cells, 9(3) (2020-03-18)
The global prevalence of type 2 diabetes (T2D) is increasing rapidly, especially in low- and middle-income countries and has a high number of associated comorbidities. Plasmatic concentrations of branched chain amino acids (BCAA) and retinol-binding protein 4 (RBP4) have been

Articles

Assay protocol for the colorimetric detection of Branched Chain Amino Acids (BCAA) in biological samples using the alanine assay kit.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service