Skip to Content
Merck
All Photos(1)

Key Documents

M9943

Sigma-Aldrich

10X MTP Taq Buffer

10X buffer, free of DNA contaminants; use with MTP Taq DNA Polymerase

Sign Into View Organizational & Contract Pricing

Select a Size

1.5 ML
$58.00

$58.00


Estimated to ship onMay 29, 2025



Select a Size

Change View
1.5 ML
$58.00

About This Item

UNSPSC Code:
41106306
NACRES:
NA.52

$58.00


Estimated to ship onMay 29, 2025


Quality Level

form

liquid

color

colorless

shipped in

wet ice

storage temp.

−20°C

General description

DNA contaminants can be introduced into PCR through a number of reagents. To minimize the risk of contaminant DNA during PCR, we offer 10x MTP Taq buffer to be used with MTP Taq DNA Polymerase (Product No. D7442). Each lot of MTP Taq buffer undergoes strict quality control testing to ensure the absence of contaminating DNA. To prevent false positive PCR results, only DNA-free reagents should be used in PCR reactions with MTP Taq DNA polymerase.

Application

10X MTPTaq Buffer has been used:

  • for the amplification of bacterial 16s rRNA genes from purified DNA [1]
  • to amplify archaeal 16s rRNA [2]
  • along with MTP Taq DNA polymerase (D7442) to amplify specific bacterial 16s rRNA genes from purified DNA [3]

Legal Information

MTP is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Camille Mazière et al.
Microbiological research, 252, 126854-126854 (2021-08-29)
In order to understand the effect of human practices on microbial mats organisation, the study aimed to investigate the biodiversity within microbial mats from exploited and abandoned salterns. Despite several attempts, archaeal 16S rRNA gene fragment sequences were not obtained
Florie Maillard et al.
Cells, 8(1) (2019-01-13)
Crohn's disease is characterized by abnormal ileal colonization by adherent-invasive E. coli (AIEC) and expansion of mesenteric adipose tissue. This study assessed the preventive effect of spontaneous physical activity (PA) on the gut-adipose tissue in a mouse model that mimics
Florie Maillard et al.
PloS one, 14(4), e0214660-e0214660 (2019-04-10)
Increased visceral adipose tissue and dysbiosis in the overweight and obese promote chronic inflammation. The aim of this study was to compare the effects of moderate-intensity continuous training (MICT) and high-intensity interval training (HIIT) on the gut-adipose tissue cross-talk in
Florie Maillard et al.
Cells, 8(1) (2019-01-13)
Crohn's disease is characterized by abnormal ileal colonization by adherent-invasive E. coli (AIEC) and expansion of mesenteric adipose tissue. This study assessed the preventive effect of spontaneous physical activity (PA) on the gut-adipose tissue in a mouse model that mimics
Florie Maillard et al.
PloS one, 14(4), e0214660-e0214660 (2019-04-10)
Increased visceral adipose tissue and dysbiosis in the overweight and obese promote chronic inflammation. The aim of this study was to compare the effects of moderate-intensity continuous training (MICT) and high-intensity interval training (HIIT) on the gut-adipose tissue cross-talk in

Protocols

MTP™ Taq DNA Polymerase is a recombinant thermostable enzyme from Thermus aquaticus expressed in E. coli and purified using a proprietary process to minimize levels of contaminating DNA.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service