Skip to Content
Merck
All Photos(1)

Key Documents

M9672

Sigma-Aldrich

6-O-α-Maltosyl-β-cyclodextrin hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C54H90O45
CAS Number:
Molecular Weight:
1459.27
MDL number:
UNSPSC Code:
12352201

form

powder

optical activity

[α]25/D 160 to 175 °, c = 0.6% (w/v) in water

storage temp.

2-8°C

General description

6-O-α-Maltosyl-β-cyclodextrin hydrate forms a soluble inclusion complex with cholesterol.

Application

6-O-α-Maltosyl-β-cyclodextrin has been used in a study to assess the effects of the cholesterol inclusion complex on cellular cholesterol levels. It has also been used in a study to investigate the enantioseparation of catechin and epicatechin.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantioseparation of catechin and epicatechin in plant food by chiral capillary electrophoresis
Kofink, M., et al.
European Food Research and Technology, 225, 569-577 (2007)
Oligomeric and functional properties of a debranching enzyme (TreX) from the archaeon Sulfolobus solfataricus P2
Park, J., et al.
Biocatalysis and Biotransformation, 26, 76-85 (2008)
Y Inoue et al.
Carbohydrate research, 226(2), 197-208 (1992-03-30)
The formation and molecular geometry of inclusion complexes of some branched cyclomaltaoses with p-nitrophenol in aqueous solution have been investigated by using high-resolution 1H-n.m.r. spectroscopy. 6-O-(alpha-Maltosyl)cyclomalto-hexaose and -heptaose were found to form 1:1 inclusion complexes with p-nitrophenol, and the dissociation
Improvement of stability and dissolution of prostaglandin E1 by maltosyl-beta-cyclodextrin in lyophilized formulation.
M Yamamoto et al.
Chemical & pharmaceutical bulletin, 40(3), 747-751 (1992-03-01)
K Matsubara et al.
Pharmaceutical research, 14(10), 1401-1405 (1997-11-14)
The present study addresses how maltosyl-beta-cyclodextrin (G2-beta-CyD) impacts upon the alpha-chymotrypsin-catalyzed hydrolysis of buserelin acetate, an agonist of luteinizing hormone-releasing hormone with emphasis upon the direct effect of G2-beta-CyD on the activity of the protease. Kinetic and solubility studies were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service