Skip to Content
Merck
All Photos(1)

Key Documents

M9635

Sigma-Aldrich

Microperoxidase (MP-9) ammonium salt from equine heart cytochrome c

~90% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C74H99FeN17O18S2
CAS Number:
Molecular Weight:
1634.66
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

Assay

~90% (HPLC)

storage temp.

−20°C

SMILES string

N.CC(C)C(NC(=O)C(NC(=O)C(Cc1cnc[nH]1)NC(=O)C2CSC(C)C3=C(C)C4=CC5=[N]6C(=Cc7c(CCC(O)=O)c(C)c8C=C9C(C(C)SCC(N)C(=O)NC(C)C(=O)NC(CCC(N)=O)C(=O)N2)=C(C)C%10=[N]9[Fe]6(N4C3=C%10)n78)C(CCC(O)=O)=C5C)C(C)O)C(=O)NC(CCC(O)=O)C(=O)NC(CCCCN)C(O)=O

InChI

1S/C74H101N17O18S2.Fe.H3N/c1-32(2)64(72(106)86-46(17-21-61(98)99)68(102)87-47(74(108)109)13-11-12-22-75)90-73(107)65(38(8)92)91-70(104)56(23-41-28-78-31-79-41)88-71(105)57-30-111-40(10)63-35(5)50-24-48-33(3)42(14-19-59(94)95)52(81-48)27-53-43(15-20-60(96)97)34(4)49(82-53)25-54-62(36(6)51(84-54)26-55(63)83-50)39(9)110-29-44(76)67(101)80-37(7)66(100)85-45(69(103)89-57)16-18-58(77)93;;/h24-28,31-32,37-40,44-47,56-57,64-65,92H,11-23,29-30,75-76H2,1-10H3,(H17,77,78,79,80,81,82,83,84,85,86,87,88,89,90,91,93,94,95,96,97,98,99,100,101,102,103,104,105,106,107,108,109);;1H3/q;+2;/p-2/b48-24-,49-25-,50-24-,51-26-,52-27-,53-27-,54-25-,55-26-;;

InChI key

SOIPPMUJPJAVPO-JTUAPZFOSA-L

Application

Of possible interest as a heme tracer in electron microscopic cytochemistry.

Biochem/physiol Actions

Has peroxidase activity.

Other Notes

The proposed structure is the heme portion of cytochrome c with amino acids 14 through 22 still attached.

Preparation Note

Prepared by enzymatic degradation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A heme-nonapeptide tracer for electron microscopy. Preparation, characterization and comparison with other heme-tracers.
H Plattner et al.
Histochemistry, 53(3), 223-242 (1977-08-22)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service