Skip to Content
Merck
All Photos(1)

Key Documents

M8447

Sigma-Aldrich

MARK1, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

KIAA1477

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

400-541 nmol/min·mg

mol wt

~125 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... MARK1(4139)

Biochem/physiol Actions

MARK1 is a member of the MARK family and is a serine/threonine-protein kinase that plays a key role in signal transduction. Due to its protein serine/threonine kinase activity, MARK1 is known to mediate phosphorylation of key proteins involved in signal transduction and cell communication. MARK1 phosphorylates microtubule-associated proteins and trigger microtubule disruption. Gene mutation studies performed in mice revealed that after targeted disruption of the MARK1 gene, the mice lacked the ability to drink, and displayed hind leg motor dysfunction.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Drewes et al.
Cell, 89(2), 297-308 (1997-04-18)
MARK phosphorylates the microtubule-associated proteins tau, MAP2, and MAP4 on their microtubule-binding domain, causing their dissociation from microtubules and increased microtubule dynamics. We describe the molecular cloning, distribution, activation mechanism, and overexpression of two MARK proteins from rat that arise
Manabu Nakayama et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 20(10), 1718-1720 (2006-06-30)
Given that thousands of genes exist in the mammalian genome, criteria are needed to prioritize their functional analysis and to decrease the likelihood of producing gene-targeted mice that lack overt phenotypes. Initial analysis efforts are likely to be fruitful if

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service