Skip to Content
Merck
All Photos(1)

Key Documents

M8273

Sigma-Aldrich

Control Ascites Fluid from murine myeloma

clone NS-1, ascites fluid, liquid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

mouse

antibody form

ascites fluid

clone

NS-1, monoclonal

form

liquid

contains

15 mM sodium azide

storage temp.

−20°C

target post-translational modification

unmodified

General description

Control Ascites Fluid is a processed mouse ascites product that is derived from the NS-1 murine myeloma cells. This myeloma cell line does not secrete immunoglobulins. The control ascites fluid has no specificity and hence it can be used as a universal control in a wide range of immunological assays. No background reactions are observed with the use of this product in immunoassays.

Application

Control Ascites Fluid can be used in immunohistochemistry[1] and gel mobility shift assays[2].
Control Ascites Fluid from murine myeloma has been used in immunocytochemistry [3] and fluorescence-activated cell sorting (FACS).[4]

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Testosterone up-regulates androgen receptors and decreases differentiation of porcine myogenic satellite cells in vitro
Doumit M, et al.
Endocrinology, 137(4), 1385-1394 (1996)
R A Levine et al.
Veterinary pathology, 39(3), 372-378 (2002-05-17)
Canine osteosarcoma (OS) cell lines contain mutations that directly or indirectly inactivate the tumor suppressor genes p53 and retinoblastoma. Another important tumor suppressor, PTEN, is mutated in many human cancers. To determine whether inactivation of PTEN plays a role in
Russell S Hamilton et al.
RNA (New York, N.Y.), 15(2), 200-207 (2009-01-16)
mRNA localization is a widespread mode of delivering proteins to their site of function. The embryonic axes in Drosophila are determined in the oocyte, through Dynein-dependent transport of gurken/TGF-alpha mRNA, containing a small localization signal that assigns its destination. A
Differential requirement for type I and type II transforming growth factor $\beta$ receptor kinase activity in ligand-mediated receptor endocytosis
Anders RA, et al.
The Journal of Biological Chemistry, 273(36), 23118-23125 (1998)
M P D'Souza et al.
AIDS (London, England), 5(9), 1061-1070 (1991-09-01)
In a National Institutes of Health (NIH)/World Health Organization (WHO)-sponsored collaboration, 26 laboratories characterized a coded panel of monoclonal antibodies (MAb) to HIV-1 envelope protein. The MAb were evaluated by serological [radioimmunoprecipitation, immunoblot, enzyme-linked immunosorbent assay (ELISA) and peptide mapping]

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service