Skip to Content
Merck
All Photos(2)

Documents

M8070

Sigma-Aldrich

Monoclonal Anti-phospho-MDMX (pTyr55) antibody produced in mouse

~2 mg/mL, clone PH-MDMX-55, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-HDM4, Anti-HDMX, Anti-MDM4, Anti-MGC132766, Anti-MRP1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

PH-MDMX-55, monoclonal

form

buffered aqueous solution

mol wt

antigen ~80 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

concentration

~2 mg/mL

technique(s)

indirect ELISA: suitable
western blot: 4-8 μg/mL using total cell extract of HEK-293T cells co-transfected with human MDMX and a specific kinase

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr55)

Gene Information

human ... MDM4(4194)

General description

Monoclonal Anti-phospho-MDMX (pTyr55) (mouse IgG2a isotype) is derived from the hybridoma PH-MDMX-55 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice. Mouse Double Minute 4 (MDM4) comprises the N-terminal p53 binding domain, a zinc finger, and an acidic region, followed by a really interesting new gene (RING) domain at C-terminus. MDM4 gene is mapped to human chromosome 1q32.1. The RING finger harbors a putative nuclear localization signal.

Specificity

Monoclonal Anti-phospho-MDMX (pTyr55) recognizes human phosphorylated MDMX (pTyr55) (approx. 80 kDa).

Immunogen

synthetic phosphopeptide corresponding to amino acids 50-64 (pTyr55) of human MDMX.

Application

Monoclonal Anti-phospho-MDMX (pTyr55) antibody produced in mouse may be used in:
  • immunoblotting
  • immunoprecipitation
  • immunocytochemistry
  • enzyme-linked immunosorbent assay (ELISA)

Biochem/physiol Actions

Mouse Double Minute 4 (MDM4 or HDMX or MDMX) undergoes phosphorylation. It is the major inhibitor of p53 transcriptional activity. Despite their significant structural similarity, mouse double minute 2 (MDM2) and MDMX play distinct roles in p53 regulation. The critical and non-redundant role of MDMX and MDM2 in the regulation of p53 in vivo is strikingly demonstrated by the lethality of the mdmx or mdm2 null embryos, which can be fully rescued by the elimination of p53. MDM2 or MDMX are frequently overexpressed in human cancers, leading to the suppression of p53. Studies on MDMX in response to stress conditions, involving post-translational modifications and protein-protein interactions are reported.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For extended storage, freeze at -20 °C in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discard if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dan Michael et al.
Current opinion in genetics & development, 12(1), 53-59 (2002-01-16)
Cells within an organism are occasionally exposed to either intracellular or environmental stress. Such stress often has genotoxic potential that enhances the probability of cancer. Two gene families, the p53 family (p53, p63 and p73) and the Mdm2 family (Mdm2
Sue Haupt et al.
Journal of molecular cell biology, 11(3), 231-244 (2019-01-29)
The mouse double minute 4 (MDM4) is emerging from the shadow of its more famous relative MDM2 and is starting to steal the limelight, largely due to its therapeutic possibilities. MDM4 is a vital regulator of the tumor suppressor p53.
Ugo Perricone et al.
MedChemComm, 9(6), 920-936 (2018-08-16)
Molecular dynamics (MD) has become increasingly popular due to the development of hardware and software solutions and the improvement in algorithms, which allowed researchers to scale up calculations in order to speed them up. MD simulations are usually used to
Orit Karni-Schmidt et al.
Annual review of pathology, 11, 617-644 (2016-03-30)
For more than 25 years, MDM2 and its homolog MDMX (also known as MDM4) have been shown to exert oncogenic activity. These two proteins are best understood as negative regulators of the p53 tumor suppressor, although they may have additional

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service