M7656
Methyl 11-methyldodecanoate
≥98% (capillary GC)
Synonym(s):
11-Methyllauric acid methyl ester
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C14H28O2
CAS Number:
Molecular Weight:
228.37
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
Assay
≥98% (capillary GC)
form
solid
functional group
ester
shipped in
ambient
storage temp.
2-8°C
SMILES string
COC(=O)CCCCCCCCCC(C)C
InChI
1S/C14H28O2/c1-13(2)11-9-7-5-4-6-8-10-12-14(15)16-3/h13H,4-12H2,1-3H3
InChI key
GSRXTORWPQDOOY-UHFFFAOYSA-N
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Gan Hui Siang et al.
Journal of chromatography. A, 1217(52), 8073-8078 (2010-11-18)
The development of a two phase hollow fiber liquid-phase microextraction technique, followed by gas-chromatography-flame ionization detection (GC-FID) for the profiling of the fatty acids (FAs) (lauric, myristic, palmitic, stearic, palmitoleic, oleic, linoleic, linolenic and arachidic) in vegetable oils is described.
Tomás Horák et al.
Journal of agricultural and food chemistry, 57(23), 11081-11085 (2009-11-13)
Solid-phase extraction (SPE), solid-phase microextraction (SPME) using carbowax/divinylbenzen fiber, and stir bar sorptive extraction (SBSE) followed by solvent back extraction have been used for the extraction of free fatty acids (caproic, caprylic, pelargonic, capric, lauric, myristic, palmitic, stearic, oleic, linoleic
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service