Skip to Content
Merck
All Photos(1)

Key Documents

M7641

Sigma-Aldrich

Metallothionein from rabbit liver

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202

biological source

rabbit liver

form

powder

Metal as Cd + Zn content

~7%

impurities

salt, essentially free

UniProt accession no.

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Other Notes

Contains both form I and form II.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiu-Juan Yu et al.
Marine pollution bulletin, 70(1-2), 274-280 (2013-03-30)
Pearl River Estuary (PRE) is the largest estuary in Southern China and there has been an increasing concern of metal pollution due to regional industrialization. In this study, we investigated the spatial variation of metal pollution (Ag, As, Cd, Cu
Wageh Sobhy Darwish et al.
The Japanese journal of veterinary research, 61 Suppl, S48-S53 (2013-05-02)
This study was undertaken to investigate the constitutive response of xenobiotic metabolizing enzymes (XMEs) to lead (Pb2+) exposure in cultured rat liver (H4IIE) cell lines. Phase I enzymes such as CYP1A1 and CYP1A2 had mRNA expressions that were slightly induced
Rezvan Mohammadi Nezhad et al.
The protein journal, 32(2), 131-137 (2013-02-07)
Metallothioneins (MTs) are ubiquitous, low molecular mass and cysteine-rich proteins that play important roles in maintaining intracellular metal homeostasis, eliminating metal toxification and protecting the cells against oxidative damages. MTs are able to bind metal ions through the thiol groups
Eva Freisinger et al.
Metal ions in life sciences, 11, 339-371 (2013-02-23)
Metallothioneins (MTs) are low-molecular-mass cysteine-rich proteins with the ability to bind mono- and divalent metal ions with the electron configuration d ( 10 ) in form of metal-thiolate clusters. MTs are thought, among others, to play a role in the
M M Sakuragui et al.
Environmental pollution (Barking, Essex : 1987), 178, 41-51 (2013-04-02)
This study investigated the relationship between contaminant body burden and the oxidative stress status of the gills and livers of two wild fish species in the Furnas Hydroelectric Power Station (HPS) reservoir (Minas Gerais, Brazil). Gills and livers presented similar

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service