Skip to Content
Merck
All Photos(1)

Key Documents

M7402

Sigma-Aldrich

Magainin II

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

0.1 MG
$301.00
0.5 MG
$1,030.00

$301.00


Estimated to ship onMarch 22, 2025



Select a Size

Change View
0.1 MG
$301.00
0.5 MG
$1,030.00

About This Item

Empirical Formula (Hill Notation):
C114H180N30O29S
CAS Number:
Molecular Weight:
2466.90
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.32

$301.00


Estimated to ship onMarch 22, 2025


Quality Level

Assay

≥97% (HPLC)

antibiotic activity spectrum

fungi
viruses

Mode of action

cell membrane | interferes

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)CN)C(=O)NCC(=O)N[C@@H](CCCCN)C(=O)N[C@@H](Cc1ccccc1)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](Cc2c[nH]cn2)C(=O)N[C@@H](CO)C(=O)N[C@@H](C)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](Cc3ccccc3)C(=O)NCC(=O)N[C@@H](CCCCN)C(=O)N[C@@H](C)C(=O)N[C@@H](Cc4ccccc4)C(=O)N[C@@H](C(C)C)C(=O)NCC(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H]([C@@H](C)CC)C(=O)N[C@@H](CCSC)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](CO)C(O)=O

InChI

1S/C114H180N30O29S/c1-12-65(7)94(142-88(148)55-119)112(170)124-59-90(150)129-74(38-24-28-45-116)100(158)137-81(51-70-33-19-15-20-34-70)106(164)135-79(49-63(3)4)105(163)138-83(53-72-56-121-62-125-72)107(165)140-85(60-145)110(168)127-67(9)96(154)131-75(39-25-29-46-117)101(159)132-76(40-26-30-47-118)102(160)136-80(50-69-31-17-14-18-32-69)98(156)122-57-89(149)128-73(37-23-27-44-115)99(157)126-68(10)97(155)134-82(52-71-35-21-16-22-36-71)109(167)143-93(64(5)6)111(169)123-58-91(151)130-77(41-42-92(152)153)104(162)144-95(66(8)13-2)113(171)133-78(43-48-174-11)103(161)139-84(54-87(120)147)108(166)141-86(61-146)114(172)173/h14-22,31-36,56,62-68,73-86,93-95,145-146H,12-13,23-30,37-55,57-61,115-119H2,1-11H3,(H2,120,147)(H,121,125)(H,122,156)(H,123,169)(H,124,170)(H,126,157)(H,127,168)(H,128,149)(H,129,150)(H,130,151)(H,131,154)(H,132,159)(H,133,171)(H,134,155)(H,135,164)(H,136,160)(H,137,158)(H,138,163)(H,139,161)(H,140,165)(H,141,166)(H,142,148)(H,143,167)(H,144,162)(H,152,153)(H,172,173)/t65-,66-,67-,68-,73-,74-,75-,76-,77-,78-,79-,80-,81-,82-,83-,84-,85-,86-,93-,94-,95-/m0/s1

InChI key

MGIUUAHJVPPFEV-ABXDCCGRSA-N

Amino Acid Sequence

Gly-Ile-Gly-Lys-Phe-Leu-His-Ser-Ala-Lys-Lys-Phe-Gly-Lys-Ala-Phe-Val-Gly-Glu-Ile-Met-Asn-Ser

General description

Chemical structure: peptide
Magainin II is a member of the antimicrobial peptides family.[1] It is a cationic peptide with Gly-Ile-Gly-Lys-Phe-Leu-His-Ser-Ala-Lys-Lys-Phe-Gly-Lys-Ala-Phe-Val-Gly-Glu-Ile-Met-Asn-Ser amino acid sequence. Magainin II is water soluble and non-hemolytic at its effective and amphiphilic antimicrobial concentrations.[2]

Biochem/physiol Actions

Antibiotic peptide. Magainins are positively charged and amphiphatic. Thought to preferentially bind to anionic phospholipids abundant in bacterial membranes with the formation of dynamic peptide-lipid supramolecular pore and cell permeabilization. Binding to artificial neutral membranes has also been demonstrated.
Magainin II acts as an antibiotic in a variety of organisms. Magainin II exhibits cytotoxic and anti-proliferative effects via pore formation in bladder cancer cells. In mice, it acts as an anti-tumor agent in several cancer cell lines and tumors.[1] Magainin II facilitates lactate dehydrogenase discharge from mammalian cells.[3] It directly interacts with certain cell membranes via amphiphilic α-helix and forms permeable ion channels, which results in depolarization and irreversible cytolysis and finally cell death. At low concentrations, Magainin II retards the growth of bacterial and fungal species and it promotes cytolysis in protozoa.[2]

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antitumor activity of the antimicrobial peptide magainin II against bladder cancer cell lines.
Lehmann J
European Urology, 50(1), 141-147 (2006)
Evan F Haney et al.
Chemistry and physics of lipids, 163(1), 82-93 (2009-10-06)
Antimicrobial peptides are naturally produced by numerous organisms including insects, plants and mammals. Their non-specific mode of action is thought to involve the transient perturbation of bacterial membranes but the molecular mechanism underlying the rearrangement of the lipid molecules to
Brijesh Kumar Pandey et al.
The Biochemical journal, 436(3), 609-620 (2011-03-26)
Cytotoxicity, a major obstacle in therapeutic application of antimicrobial peptides, is controlled by leucine-zipper-like sequences in melittin and other naturally occurring antimicrobial peptides. Magainin 2 shows significantly lower cytotoxicity than many naturally occurring antimicrobial peptides and lacks this structural element.
Mikyung Han et al.
Biophysical journal, 97(1), 164-172 (2009-07-08)
Magainin, a 23-residue antibiotic peptide, interacts directly with the lipid bilayer leading to cell lysis in a strongly concentration-dependent fashion. Utilizing cryo-electron microscopy, we have directly observed magainin interacting with synthetic DMPC/DMPG membranes. Visual examination shows that visibly unperturbed vesicles
The cytotoxic effect of magainin II on the MDA-MB-231 and M14K tumour cell lines.
Anghel R
BioMed Research International, 2013:831709 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service