Skip to Content
Merck
All Photos(1)

Key Documents

M6895

Sigma-Aldrich

MEM Vitamin Solution (100×)

sterile-filtered, BioReagent, suitable for cell culture

Synonym(s):

MEM media, MEM vitamin solution

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$67.90

$67.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$67.90

About This Item

UNSPSC Code:
12352205
eCl@ss:
32160406
NACRES:
NA.75

$67.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

sterility

sterile-filtered

Quality Level

product line

BioReagent

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

color

yellow to dark yellow

pH

7.0-8.0

application(s)

agriculture

shipped in

dry ice

storage temp.

−20°C

General description

MEM Vitamin Solution (100×) is a sterile-filtered bioreagent, which is suitable for cell culture.
Synthetic/ basal media used currently contain amino acids, carbohydrates, vitamins and salts. Eagle′s minimum essential medium (MEM) is a modified version of BME (basal medium Eagle′s), which contains increased amounts of each amino acid.[1] Addition of water soluble vitamins such as, inositol to various culture facilitates the hatching of hamster and rabbit blastocysts. Vitamins also influence the metabolism of glucose in sheep and mouse embryos.[2]

Application

MEM Vitamin Solution (100×) has been used in:
  • glial conditioned medium for primary neuronal culture[3]
  • in Luria-Bertani (LB) medium or minimal medium (MM)[4]
  • in private medium (PM = aminoacid-starved medium) for the Aa deprivation experiments[5]

MEM Vitamin Solution (100×) is used for the following applications:
  • In the preparation of DMEM medium (for primary neural cell culture)[6][3][7]
  • For the maintenance of cell lines in Eagle′s Minimum Essential Medium (the medium is supplemented with MEM Vitamin Solution along with other components)[8]

Adding supplements of vitamins to media stimulates growth and prolongs the viability of cells in culture. This concentrated supplement adds nutrients and reduces the biosynthetic burden on cell in vitro.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paradoxical role of the NADPH oxidase NOX4 in early preneoplastic stages of hepatocytes induced by amino acid deprivation
Campisano S, et al.
Biochimica et biophysica acta. General subjects, 1863(4), 714-722 (2019)
Motoko Tanaka et al.
The Kobe journal of medical sciences, 57(3), E116-E127 (2011-01-01)
Although the viral factors of host adaptation from domestic poultry to humans have been studied several times since the first cases of direct transmission of highly pathogenic avian influenza viruses from domestic poultry to humans were confirmed in 1997, the
Kenneth Verstraete et al.
Nature communications, 8, 14937-14937 (2017-04-04)
The pro-inflammatory cytokine thymic stromal lymphopoietin (TSLP) is pivotal to the pathophysiology of widespread allergic diseases mediated by type 2 helper T cell (Th2) responses, including asthma and atopic dermatitis. The emergence of human TSLP as a clinical target against
Culture Media, Solutions, and Systems in Human ART
Quinn P
Culture Media, Solutions, and Systems in Human ART. Cambridge University Press (2014)
Ken-Ichi Oinuma et al.
Journal of bacteriology, 197(6), 1115-1124 (2015-01-15)
The degradation mechanisms of natural and artificial hydrazides have been elucidated. Here we screened and isolated bacteria that utilize the acylhydrazide 4-hydroxybenzoic acid 1-phenylethylidene hydrazide (HBPH) from soils. Physiological and phylogenetic studies identified one bacterium as Microbacterium sp. strain HM58-2

Questions

  1. MEM ビタミン溶液(100X)の組成を教えていただきたいです。

    1 answer
    1. The MEM vitamin solution is a published classical formulation containing numerous components. See the link below to review the formulation of this and other classical vitamin and amino acid mixtures:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/167/485/m6895for.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service