Skip to Content
Merck
All Photos(1)

Documents

M6807

Sigma-Aldrich

Misoprostol

≥99% (HPLC)

Synonym(s):

(±)-15-Deoxy-(16RS)-16-hydroxy-16-methylprostaglandin E1 methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H38O5
CAS Number:
Molecular Weight:
382.53
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥99% (HPLC)

form

liquid

functional group

ester

storage temp.

−20°C

SMILES string

CCCCC(C)(O)C\C=C\[C@H]1[C@H](O)CC(=O)[C@@H]1CCCCCCC(=O)OC

InChI

1S/C22H38O5/c1-4-5-14-22(2,26)15-10-12-18-17(19(23)16-20(18)24)11-8-6-7-9-13-21(25)27-3/h10,12,17-18,20,24,26H,4-9,11,13-16H2,1-3H3/b12-10+/t17-,18-,20-,22?/m1/s1

InChI key

OJLOPKGSLYJEMD-URPKTTJQSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

PGE1 analog prodrug which is rapidly de-esterified to active "misoprostolic acid". Cited for extremely wide-ranging therapeutic effects, including prevention of NSAID-induced gastric ulceration, regulation of immunologic cascades, inhibition of platelet-activating factor (PAF), treatment of ethanol- and acetaminophen-induced hepatotoxicity and hepatitis, and stimulation of cartilage repair after injury.
Cytoprotective PGE1 analog that prevents NSAID-induced gastric ulceration.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ingrid Sääv et al.
Human reproduction (Oxford, England), 30(6), 1314-1322 (2015-04-04)
Can sublingual administration of misoprostol 1 h prior to vacuum aspiration be more effective than vaginal administration and as effective as either route three 3 h prior to surgery? Sublingually administered misoprostol is superior to vaginally administered misoprostol when given
Christina S Chu et al.
Journal of the Royal Society of Medicine, 105(8), 336-347 (2012-08-22)
This article describes and critically appraises clinical trials assessing misoprostol effectiveness in preventing primary postpartum haemorrhage (PPH) in home and community settings in low- and middle-income countries. Of 172 identified studies of misoprostol use in labour only six fulfilled the
V A Hundley et al.
BJOG : an international journal of obstetrics and gynaecology, 120(3), 277-285 (2012-11-30)
Using misoprostol to prevent postpartum haemorrhage (PPH) in home-birth settings remains controversial. To review the safety and effectiveness of oral misoprostol in preventing PPH in home-birth settings. The Cochrane Library, PubMed, and POPLINE were searched for articles published until 31
D Y Graham et al.
Lancet (London, England), 2(8623), 1277-1280 (1988-12-03)
A double-blind, placebo-controlled study was carried out to see whether the synthetic E prostaglandin, misoprostol, would prevent gastric ulcer induced by non-steroidal anti-inflammatory drugs (NSAIDs). 420 patients with osteoarthritis and NSAID-associated abdominal pain were studied; they were receiving ibuprofen, piroxicam
N Bernard et al.
BJOG : an international journal of obstetrics and gynaecology, 120(5), 568-574 (2013-01-26)
To report the follow-up of continuing pregnancies after first-trimester exposure to mifepristone. Observational prospective study. France. Patients exposed to mifepristone during the first 12 weeks of pregnancy. Women were included in the study when they or their doctors asked a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service