Skip to Content
Merck
All Photos(1)

Documents

M5686

Greiner high and medium binding 96 well plates

flat bottom clear polystyrene wells (with micro-clear bottom), medium binding surface, non-sterile

Synonym(s):

96 multiwell plates, 96 well microplates, 96 well microtiter plates, 96 well strip plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
NACRES:
NB.22

material

black polystyrene
flat bottom clear polystyrene wells (with micro-clear bottom)

sterility

non-sterile

feature

lid: no
skirt (F-bottom)

packaging

case of 40

manufacturer/tradename

Greiner 655096

maximum volume

392 μL

size

96 wells

working volume

340 μL

binding type

medium binding surface

Looking for similar products? Visit Product Comparison Guide

General description

FLUOTRAC 200 96W Microplate, well working volume 25-340 μL; total well volume 392 μL.
Polystyrene Multiwell plates from Greiner for immunoassay and high throughput screening applications. High binding plates are recommended for ELISAs. High binding denotes a binding capacity of 600 ng/cm2 and scatter of less than 10%. Optimal protein binding is variable and comparative tests between high and medium binding plates is suggested.

Other Notes

96 well flat (F) μClear® bottom; medium binding microplate, chimney style, no lid, alphanumeric well coding, length: 127.76 mm; width: 85.48 mm; curvature approx. 200 um.

Suitability

Black plates minimize background fluorescence. FLUOTRAC 200 binds up to 200 ng/cm2. Clear bottoms allow for monitoring thru the plate bottom. Free of detectable DNase/RNase, human DNA and pyrogens.

Legal Information

μClear is a registered trademark of Greiner Bio-One GmbH
FLUOTRAC is a trademark of Greiner Bio-One GmbH

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

This assay protocol is suitable for the colorimetric/fluorometric detection of phenylalanine in cell and tissue culture supernatants, urine, plasma, serum, and other biological samples using the Phenylalanine Assay Kit (MAK005). Phenylalanine concentration is determined by a coupled enzyme assay, which results in the deamination of phenylalanine and the production of NADH which reacts with the probe resulting in a fluorometric (λex = 535/λem = 587 nm) product, proportional to the phenylalanine present. The linear range of detection for this assay is between 0.2–1.0 nmole.

Assay protocol for the colorimetric/fluorometric detection of Alanine in biological samples using the alanine assay kit.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service