Skip to Content
Merck
All Photos(1)

Key Documents

M5046

Sigma-Aldrich

(±)-α-Methyl-(4-sulfonophenyl)glycine

Synonym(s):

MSPG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11NO5S
CAS Number:
Molecular Weight:
245.25
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

storage temp.

2-8°C

SMILES string

CC(N)(C(O)=O)c1ccc(cc1)S(O)(=O)=O

InChI

1S/C9H11NO5S/c1-9(10,8(11)12)6-2-4-7(5-3-6)16(13,14)15/h2-5H,10H2,1H3,(H,11,12)(H,13,14,15)

InChI key

MVDSFPIEJILRME-UHFFFAOYSA-N

Biochem/physiol Actions

Antagonist at Group III metabotropic glutamate receptors.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ivana Pulić et al.
Journal of structural biology, 194(2), 147-155 (2016-02-13)
Flagellin component D (FlgD) participates in the assembly of flagella, helical tubular structures that provide motility in non-filamentous bacteria. FlgD guides and controls the polymerization of FlgE that builds the hook, a short curved and hollow cylinder that connects the
Ersin Yavas et al.
ACS chemical neuroscience, 8(2), 320-328 (2017-01-26)
The N-methyl-d-aspartate (NMDA) receptor antagonist, phencyclidine, induces behavioral changes in rodents mimicking symptoms of schizophrenia, possibly mediated through dysregulation of glutamatergic control of mesolimbic dopamine release. We tested the hypothesis that NMDA receptor activation modulates accumbens dopamine release, and that
D E Jane et al.
Neuropharmacology, 34(8), 851-856 (1995-08-01)
The depression of the monosynaptic excitation of neonatal rat motoneurones produced by the metabotropic glutamate receptor (mGluR) agonists (1S,3S)-1-aminocyclopentane-1, 3-dicarboxylate (ACPD) or L-2-amino-4-phosphonobutyrate (L-AP4) was antagonized by three novel phenylglycine analogues: (RS)-alpha-methyl-4-sulphonophenylglycine (MSPG), (RS)-alpha-methyl-4-phosphonophenylglycine (MPPG) and (RS)-alpha-methyl-4-tetrazolylphenylglycine (MTPG). The potencies
Lorenzo Caputi et al.
Nature chemical biology, 16(4), 383-386 (2020-02-19)
Cycloaddition reactions generate chemical complexity in a single step. Here we report the crystal structures of three homologous plant-derived cyclases involved in the biosynthesis of iboga and aspidosperma alkaloids. These enzymes act on the same substrate, named angryline, to generate
J S Bedingfield et al.
European journal of pharmacology, 309(1), 71-78 (1996-08-01)
The metabotropic glutamate (mGlu) receptor antagonist properties of novel phenylglycine analogues were investigated in adult rat cortical slices (mGlu receptors negatively coupled to adenylyl cyclase), neonatal rat cortical slices and in cultured rat cerebellar granule cells (mGlu receptors coupled to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service