Skip to Content
Merck
All Photos(3)

Key Documents

M4758

Sigma-Aldrich

(±)-6-Methyl-5,6,7,8-tetrahydropterine dihydrochloride

~95% (TLC)

Synonym(s):

6-MPH4, DL-2-Amino-4-hydroxy-6-methyl-5,6,7,8-tetrahydropteridine dihydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$267.00
100 MG
$475.00

$267.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025



Select a Size

Change View
50 MG
$267.00
100 MG
$475.00

About This Item

Empirical Formula (Hill Notation):
C7H11N5O · 2HCl
CAS Number:
Molecular Weight:
254.12
Beilstein:
5648114
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.51

$267.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Assay

~95% (TLC)

Quality Level

storage temp.

−20°C

SMILES string

Cl.Cl.CC1CNc2nc(N)nc(O)c2N1

InChI

1S/C7H11N5O.2ClH/c1-3-2-9-5-4(10-3)6(13)12-7(8)11-5;;/h3,10H,2H2,1H3,(H4,8,9,11,12,13);2*1H

InChI key

MKQLORLCFAZASZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

6-MPH4 is used to study mechanisms of nitric oxide (NO) synthase and free radical induced L-DOPA release from striatal tissue.

Biochem/physiol Actions

Synthetic cofactor for tyrosine hydrolase; also cofactor for phenylalanine and tryptophan hydroxylases; less activity than the natural cofactor, BH4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

M Bauer et al.
Journal of neurochemistry, 82(5), 1300-1310 (2002-10-03)
Glial cell line-derived neurotrophic factor (GDNF) protects dopaminergic neurones against toxic and physical damage. In addition, GDNF promotes differentiation and structural integrity of dopaminergic neurones. Here we show that GDNF can support the function of primary dopaminergic neurones by triggering
Elizabeth A Gaskell et al.
PloS one, 4(3), e4801-e4801 (2009-03-12)
The genome of the protozoan parasite Toxoplasma gondii was found to contain two genes encoding tyrosine hydroxylase; that produces L-DOPA. The encoded enzymes metabolize phenylalanine as well as tyrosine with substrate preference for tyrosine. Thus the enzymes catabolize phenylalanine to
P Abreu-González et al.
European journal of pharmacology, 541(1-2), 33-37 (2006-06-06)
In the present study we have analyzed the effect of tetrahydrobiopterin (BH4) essential cofactor for tyrosine hydroxylase and nitric oxide synthase, on the 3,4-dihydroxyphenylalanine (L-DOPA) release from in vitro incubated striatal tissue. dl-6-methyl-5,6,7,8 tetrahydropterine (6-MPH4)-stimulated L-DOPA release in a concentration-dependent
M Naoi et al.
Journal of chromatography, 427(2), 229-238 (1988-06-03)
A simple assay procedure for tyrosine hydroxylase activity in crude tissue samples was devised that requires minimal sample preparation and use of high-performance liquid chromatography with coulometric electrochemical detection. After incubation of enzyme samples, such as human brain homogenates or
F García-Molina et al.
Biochimica et biophysica acta, 1794(12), 1766-1774 (2009-08-22)
There is controversy in the literature concerning the action of tetrahydropterines on the enzyme tyrosinase and on melanogenesis in general. In this study, we demonstrate that tetrahydropterines can inhibit melanogenesis in several ways: i) by non-enzymatic inhibition involving purely chemical

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service