Skip to Content
Merck
All Photos(1)

Key Documents

M4542

Sigma-Aldrich

Melanin Concentrating Hormone rat

≥90% (HPLC)

Synonym(s):

MCH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C105H160N30O26S4
CAS Number:
Molecular Weight:
2386.84
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.32

biological source

synthetic (organic)

Assay

≥90% (HPLC)

form

powder

composition

Peptide content, ≥70%

solubility

water: 1 mg/mL, clear, colorless

UniProt accession no.

shipped in

ambient

storage temp.

−20°C

SMILES string

CSCC[C@@H]1NC(=O)[C@H](CSSC[C@H](NC(=O)[C@@H]2CCCN2C(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](Cc3ccc(O)cc3)NC(=O)[C@@H](NC(=O)[C@H](CCCNC(N)=N)NC(=O)CNC(=O)[C@H](CC(C)C)NC1=O)C(C)C)C(=O)N[C@@H](Cc4c[nH]c5ccccc45)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](C(C)C)C(O)=O)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](CCSC)NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](Cc6ccccc6)NC(=O)[C@@H](N)CC(O)=O

InChI

1S/C105H160N30O26S4/c1-53(2)42-70-86(144)118-50-80(138)119-64(24-16-36-114-103(108)109)90(148)133-83(55(5)6)100(158)130-73(45-58-28-30-60(136)31-29-58)93(151)124-69(26-18-38-116-105(112)113)101(159)135-39-19-27-78(135)99(157)132-77(98(156)128-74(46-59-49-117-63-23-15-14-22-61(59)63)95(153)121-66(32-33-79(107)137)91(149)134-84(56(7)8)102(160)161)52-165-164-51-76(97(155)123-68(35-41-163-10)88(146)126-70)131-87(145)65(25-17-37-115-104(110)111)120-92(150)71(43-54(3)4)127-89(147)67(34-40-162-9)122-96(154)75(48-82(141)142)129-94(152)72(44-57-20-12-11-13-21-57)125-85(143)62(106)47-81(139)140/h11-15,20-23,28-31,49,53-56,62,64-78,83-84,117,136H,16-19,24-27,32-48,50-52,106H2,1-10H3,(H2,107,137)(H,118,144)(H,119,138)(H,120,150)(H,121,153)(H,122,154)(H,123,155)(H,124,151)(H,125,143)(H,126,146)(H,127,147)(H,128,156)(H,129,152)(H,130,158)(H,131,145)(H,132,157)(H,133,148)(H,134,149)(H,139,140)(H,141,142)(H,160,161)(H4,108,109,114)(H4,110,111,115)(H4,112,113,116)/t62-,64-,65-,66-,67-,68-,69-,70-,71-,72-,73-,74-,75-,76-,77-,78-,83-,84-/m0/s1

InChI key

MWLPXSMFEPHETN-QRXCLJFASA-N

Gene Information

rat ... Pmch(24659)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenting Cheng et al.
Talanta, 219, 121242-121242 (2020-09-06)
Growing evidence suggests that exosomes-encapsulated miRNAs detection is of paramount significance in early diagnostics of cancer due to protection from degradation by ribonuclease. However, exosomal microRNAs with low abundance and subtle variation have restricted their clinical application. Herein, an electrochemical
Melanin-concentrating hormone induces insulin resistance through a mechanism independent of body weight gain
Marcio P
The Journal of Endocrinology (2005)
The role of melanin concentrating hormone (MCH) in the central chemoreflex: a knockdown study by siRNA in the lateral hypothalamus in rats.
Li N
PLoS ONE (2014)
Peng Miao et al.
Small (Weinheim an der Bergstrasse, Germany), 16(47), e2004518-e2004518 (2020-11-04)
miRNAs, a class of endogenous noncoding RNAs, are involved in many crucial biological processes, which have emerged as a new set of biomarkers for disease theranostics. Exploring efficient signal amplification strategy is highly desired to pursue a highly sensitive miRNA
Melanin concentrating hormone induces hippocampal acetylcholine release via the medial septum in rats.
Peptides (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service