Skip to Content
Merck
All Photos(1)

Key Documents

M4264

Sigma-Aldrich

Morphiceptin hydrochloride

≥97% (HPLC)

Synonym(s):

β-Casomorphin 1-4 amide, Tyr-Pro-Phe-Pro-NH2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H35N5O5
CAS Number:
Molecular Weight:
521.61
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (HPLC)

storage temp.

−20°C

SMILES string

Cl.NC(Cc1ccc(O)cc1)C(=O)N2CCCC2C(=O)NC(Cc3ccccc3)C(=O)N4CCCC4C(N)=O

InChI

1S/C28H35N5O5.ClH/c29-21(16-19-10-12-20(34)13-11-19)27(37)33-15-5-9-24(33)26(36)31-22(17-18-6-2-1-3-7-18)28(38)32-14-4-8-23(32)25(30)35;/h1-3,6-7,10-13,21-24,34H,4-5,8-9,14-17,29H2,(H2,30,35)(H,31,36);1H

InChI key

ARSDMYCQCLQDBN-UHFFFAOYSA-N

Biochem/physiol Actions

Agonist for μ opioid receptors

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jakub Fichna et al.
Biochemical pharmacology, 69(1), 179-185 (2004-12-14)
In the present study we investigated and compared the in vivo analgesia of centrally administered endomorphin-2 and morphiceptin, and their analogs modified in position 3. Two series of analogs were synthesized by introducing unnatural aromatic amino acids in the D
Jakub Fichna et al.
The Journal of pharmacology and experimental therapeutics, 317(3), 1150-1154 (2006-02-25)
A functional assay, based on aequorin-derived luminescence triggered by receptor-mediated changes in intracellular calcium levels, was used to examine relative potency and efficacy of the mu-opioid agonists endomorphin-1, endomorphin-2, morphiceptin, and their position 3-substituted analogs, as well as the delta-agonist
K S Rózsa et al.
General pharmacology, 27(8), 1337-1345 (1996-12-01)
1. The interaction between GABA and opioid peptides (met-enkephalin and morphiceptin) was studied on the identified, isolated and internally perfused neurons of Lymnaea stagnalis L. (Gastropoda, Basommatophora). 2. GABA (10(-7)-10(-5)M) activated a Cl-dependent inward current with about -20 mV equilibrium
Toru Ota et al.
Journal of agricultural and food chemistry, 53(15), 6112-6116 (2005-07-21)
Morphiceptin (Tyr-Pro-Phe-Pro-NH(2)), tetrapeptide, was synthesized using dipeptidyl aminopeptidase IV (DP IV, EC 3.4.14.5) derived from Aspergillus oryzae RIB 915 as a catalyst. Tyr-Pro-OEt was incubated with Phe-Pro-NH(2) in the presence of DP IV under various conditions of temperature, concentrations of
Anna Janecka et al.
Biochemical pharmacology, 71(1-2), 188-195 (2005-11-18)
Cyclic analogs of the opioid peptides endomorphin-2 and morphiceptin of the type Tyr-X-Phe-Phe-Y-NH2 and Tyr-X-Phe-D-Pro-Y-NH2 (X = Lys or Asp and Y = Lys or Asp), respectively, were synthesized in order to test their structure-activity relationships. Antinociceptive activity of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service