Skip to Content
Merck
All Photos(1)

Key Documents

M4199

Sigma-Aldrich

Myelin Oligodendrocyte Glycoprotein (40-54), Rat, Mouse

lyophilized powder

Synonym(s):

MOG (40-54)

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
$150.00
1 MG
$256.00

$150.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 μG
$150.00
1 MG
$256.00

About This Item

Empirical Formula (Hill Notation):
C84H127N27O21
CAS Number:
Molecular Weight:
1851.08
UNSPSC Code:
12352202
NACRES:
NA.32

$150.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

biological source

rat

Quality Level

Assay

≥97% (HPLC)

form

lyophilized powder

color

white

solubility

deionized water: soluble

UniProt accession no.

storage temp.

−20°C

Gene Information

mouse ... Mog(17441)
rat ... Mog(24558)

Related Categories

Amino Acid Sequence

Tyr-Arg-Ser-Pro-Phe-Ser-Arg-Val-Val-His-Leu-Tyr-Arg-Asn-Gly

Biochem/physiol Actions

The 21-residue peptide MOG 35-55 contains an antigenic epitope responsible for the activation of CD8 encephalitogenic T cells in mice. Two truncated peptides, MOG 40-54 and MOG 44-54 were shown to bind the recombinant H-2Db protein. This complex then binds MOG specific CD8 T cells. While MOG 40-54 retained the full capability of inducing paralytic disease, MOG 44-54 only partialy causes clinical paralysis and central nervous system inflamation.

Physical form

Lyophilized from 0.1% aqueous TFA.

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel pathogenic epitopes of myelin oligodendrocyte glycoprotein induce experimental autoimmune encephalomyelitis in C57BL/6 mice
Cecile Delarasse
Immunology (2013)
Oligodendrocyte myelin glycoprotein (OMgp) in rat hippocampus is depleted by chronic ethanol consumption.
Okamoto H
Neuroscience Letters (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service