Skip to Content
Merck
All Photos(1)

Key Documents

M3814

Sigma-Aldrich

Myosin light chain kinase substrate (skeletal muscle) trifluoroacetate salt

≥90% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200

Assay

≥90% (HPLC)

mol wt

calculated mol wt 1460.8

composition

Peptide content, ≥80%

solubility

H2O: soluble

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... MYL9(10398)

Amino Acid Sequence

Ala-Lys-Arg-Pro-Gln-Arg-Ala-Thr-Ser-Asn-Val-Phe-Ser-NH2

Other Notes

Myosin light chain kinase (MLCK) substrate specific for rabbit skeletal muscle

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service