Skip to Content
Merck
All Photos(1)

Key Documents

M3574

Sigma-Aldrich

Methylone hydrochloride

≥98% (HPLC)

Synonym(s):

2-Methylamino-3,4-methylene-dioxy-propiophenone hydrochloride, Methylenedioxymethcathinone hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13NO3·HCl
CAS Number:
Molecular Weight:
243.69
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

drug control

USDEA Schedule I; psicótropo (Spain)

storage condition

desiccated

color

off-white to light yellow

solubility

H2O: ≥30 mg/mL

SMILES string

Cl.CNC(C)C(=O)c1ccc2OCOc2c1

InChI

1S/C11H13NO3.ClH/c1-7(12-2)11(13)8-3-4-9-10(5-8)15-6-14-9;/h3-5,7,12H,6H2,1-2H3;1H

InChI key

GASYWEXOTOXXLK-UHFFFAOYSA-N

Biochem/physiol Actions

Methylone is a stimulant, an analog of Methamphetamine. Triple reuptake inhibitor; inhibits the re-uptake of dopamine, 5-HT and norepinephrine.
Methylone is an analog of canthinone and methamphetamine analog.

Features and Benefits

This compound is featured on the Biogenic Amine Transporters page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service