Skip to Content
Merck
All Photos(2)

Key Documents

M3184

Sigma-Aldrich

MG 624

≥98%

Synonym(s):

N,N,N-Triethyl-2-(4-trans-stilbenoxy)ethylammonium iodide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$166.00
25 MG
$524.00

$166.00


Estimated to ship onMarch 31, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$166.00
25 MG
$524.00

About This Item

Empirical Formula (Hill Notation):
C22H30INO
CAS Number:
Molecular Weight:
451.38
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$166.00


Estimated to ship onMarch 31, 2025


Request a Bulk Order

Quality Level

Assay

≥98%

solubility

DMSO: ≤22 mg/mL
H2O: insoluble

storage temp.

2-8°C

SMILES string

[I-].CC[N+](CC)(CC)CCOc1ccc(cc1)\C=C\c2ccccc2

InChI

1S/C22H30NO.HI/c1-4-23(5-2,6-3)18-19-24-22-16-14-21(15-17-22)13-12-20-10-8-7-9-11-20;/h7-17H,4-6,18-19H2,1-3H3;1H/q+1;/p-1/b13-12+;

InChI key

RDTKUZXIHMTSJO-UEIGIMKUSA-M

Gene Information

human ... CHRNA7(1139)

Biochem/physiol Actions

Nicotinic acetylcholine receptor antagonist; selectively inhibits neuronal nicotinic α-bungarotoxin sensitive receptors that contain the α7 subunit.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Gotti et al.
British journal of pharmacology, 124(6), 1197-1206 (1998-08-28)
1. Starting from the structure of an old 4-oxystilbene derivate with ganglioplegic activity (MG624), we synthesized two further derivates (F2 and F3) and two stereoisomers of F3 (F3A and F3B), and studied their selective effect on neuronal nicotinic acetylcholine receptor
Glenn W Vicary et al.
Respiratory research, 18(1), 115-115 (2017-06-04)
Tobacco-related chronic lung diseases are characterized by alterations in lung architecture leading to decreased lung function. Knowledge of the exact mechanisms involved in tobacco-induced tissue remodeling and inflammation remains incomplete. We hypothesize that nicotine stimulates the expression of extracellular matrix

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service