Skip to Content
Merck
All Photos(1)

Key Documents

M2137

Sigma-Aldrich

N-Methyl-DL-aspartic acid

Synonym(s):

(±)-2-(Methylamino)succinic acid, NMA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCOCH2CH(NHCH3)COOH
CAS Number:
Molecular Weight:
147.13
Beilstein:
1724430
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (TLC)

form

powder

color

white

SMILES string

CNC(CC(O)=O)C(O)=O

InChI

1S/C5H9NO4/c1-6-3(5(9)10)2-4(7)8/h3,6H,2H2,1H3,(H,7,8)(H,9,10)

InChI key

HOKKHZGPKSLGJE-UHFFFAOYSA-N

Biochem/physiol Actions

N-Methyl-DL-aspartic acid is a derivative of aspartic acid.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chris H P van den Akker et al.
Journal of pediatric gastroenterology and nutrition, 70(5), 664-680 (2020-04-26)
More than 10,000 preterm infants have participated in randomised controlled trials on probiotics worldwide, suggesting that probiotics in general could reduce rates of necrotising enterocolitis (NEC), sepsis, and mortality. Answers to relevant clinical questions as to which strain to use
Akin Cayir et al.
Biomarkers : biochemical indicators of exposure, response, and susceptibility to chemicals, 24(6), 592-599 (2019-04-26)
Purpose: The main goal of the present study was to determine DNA damage in pesticide-exposed greenhouse workers and pesticides non-exposed controls. Materials and methods: The DNA damage was measured by alkaline comet assay method (pH > 13) in 41 greenhouse workers and
Andrea C Tricco et al.
BMC medicine, 14(1), 216-216 (2016-12-23)
Although serotonin (5-HT3) receptor antagonists are effective in reducing nausea and vomiting, they may be associated with increased cardiac risk. Our objective was to examine the comparative safety and effectiveness of 5-HT3 receptor antagonists (e.g., dolasetron, granisetron, ondansetron, palonosetron, tropisetron)
B Häggman-Henrikson et al.
Journal of oral rehabilitation, 44(10), 800-826 (2017-09-09)
This health technology assessment evaluated the efficacy of pharmacological treatment in patients with oro-facial pain. Randomised controlled trials were included if they reported pharmacological treatment in patients ≥18 years with chronic (≥3 months) oro-facial pain. Patients were divided into subgroups:
Yan Luo et al.
BMJ open, 8(12), e023222-e023222 (2018-12-12)
Depressive disorders are the most common, burdensome and costly mental disorders. Their treatments have developed through the past decades and we now have more than a dozen new generation antidepressants, while a series of guidelines have been published to provide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service