Skip to Content
Merck
All Photos(1)

Documents

M1818

Sigma-Aldrich

Molindone hydrochloride

≥98% (HPLC), solid

Synonym(s):

3-Ethyl-1,5,6,7-tetrahydro-2-methyl-5-(4-morpholinylmethyl)-4H-indol-4-one hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H24N2O2 · HCl
CAS Number:
Molecular Weight:
312.83
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (HPLC)

form

solid

color

off-white

solubility

H2O: 19 mg/mL

originator

Endo

SMILES string

CCC1=C(C)NC2=C1C(C(CN3CCOCC3)CC2)=O.Cl

InChI

1S/C16H24N2O2.ClH/c1-3-13-11(2)17-14-5-4-12(16(19)15(13)14)10-18-6-8-20-9-7-18;/h12,17H,3-10H2,1-2H3;1H

InChI key

GQWNECFJGBQMBO-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

D2 dopamine receptor antagonist; MAO inhibitor.

Features and Benefits

This compound was developed by Endo. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aripiprazole-induced agitation after clozapine discontinuation: a case report.
Dara Y Cho et al.
The Journal of clinical psychiatry, 70(1), 141-143 (2009-02-19)
H B Pinkofsky
Annals of clinical psychiatry : official journal of the American Academy of Clinical Psychiatrists, 9(3), 175-179 (1997-10-27)
The onset of psychosis during pregnancy presents several difficult management decisions and a careful risk-benefit analysis is required. Withholding antipsychotic treatment may produce more risks than benefits. Studies on neuroleptic teratogenicity are contradictory. Most of the commonly used neuroleptics exhibit
Robert L Findling et al.
Journal of the American Academy of Child and Adolescent Psychiatry, 49(6), 583-594 (2010-05-25)
To examine the long-term safety and efficacy of three antipsychotics in early-onset schizophrenia spectrum disorders. Patients (8 to 19 years old) who had improved during an 8-week, randomized, double-blind acute trial of olanzapine, risperidone, or molindone (plus benztropine) were eligible
T Baptista et al.
Pharmacopsychiatry, 35(6), 205-219 (2003-01-09)
Excessive body weight gain (BWG) is a common side effect of some typical and atypical antipsychotic drugs (APs). Convergent evidence suggests a hierarchy in the magnitude of BWG that may be induced by diverse agents, being very high for clozapine
Unilateral auditory hallucinations in a boy with ipsilateral conductive hearing loss.
J R Brasic et al.
Journal of neurology, neurosurgery, and psychiatry, 62(3), 302-302 (1997-03-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service