Skip to Content
Merck
All Photos(1)

Key Documents

M1514

Sigma-Aldrich

Nω-Methyl-5-hydroxy­trypt­amine oxalate salt

≥90% (HPLC)

Synonym(s):

Nω-Methylserotonin oxalate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14N2O · C2H2O4
CAS Number:
Molecular Weight:
280.28
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.77

Assay

≥90% (HPLC)

storage temp.

2-8°C

SMILES string

OC(=O)C(O)=O.CNCCc1c[nH]c2ccc(O)cc12

InChI

1S/C11H14N2O.C2H2O4/c1-12-5-4-8-7-13-11-3-2-9(14)6-10(8)11;3-1(4)2(5)6/h2-3,6-7,12-14H,4-5H2,1H3;(H,3,4)(H,5,6)

InChI key

DYOZWAJOUTVNAF-UHFFFAOYSA-N

Biochem/physiol Actions

Serotonin receptor ligand.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Niels Hadrup et al.
Drug and chemical toxicology, 42(1), 76-83 (2018-07-24)
Selenium (Se) nanoparticles have been proposed as food supplements. However, the particle formulation may exert unexpected toxicity. The aim was therefore to compare toxicity of low doses of Se nanoparticles and the dissolved, ionized Se species selenite. Female rats were
A R Johnston et al.
Experimental brain research, 93(2), 293-298 (1993-01-01)
The effects of 5-hydroxytryptamine (5-HT) and related compounds on the discharge rate of tonically active medial vestibular nucleus (MVN) neurones were studied in an in vitro slice preparation of the dorsal brainstem of the rat. The majority (87 of 107
D Alberghina et al.
Research in veterinary science, 90(3), 392-395 (2010-07-17)
To evaluate the changes in plasma and platelet serotonin (5-HT) as markers of the serotoninergic system in equines, 5-HT content was measured by high performance liquid chromatography (HPLC) in deproteinized plasma obtained from peripheral blood samples of 12 clinically healthy
G L Laekeman et al.
Agents and actions, 16(5), 443-445 (1985-07-01)
Pharmacological effects of 5-hydroxykynuramine (5-HK) were investigated in chicken whole blood. It was shown that 5-HK had a time-depending inhibiting activity on platelet aggregation induced by 5-hydroxytryptamine (5-HT). Experiments were carried out in order to determine whether the inhibitory effect
Emiko Yanase et al.
Bioscience, biotechnology, and biochemistry, 74(9), 1951-1952 (2010-09-14)
Two serotonin derivatives, N,N-dimethylserotonin 5-O-β-glucoside (1a) and N-methylserotonin 5-O-β-glucoside (1b) were isolated from immature seeds of Zanthoxylum piperitum. Their structures were determined by multi-step conversion reactions and spectroscopic analyses. Immature seeds of Z. piperitum contained extremely high levels of compounds

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service