Skip to Content
Merck
All Photos(1)

Key Documents

M1445

Sigma-Aldrich

Anti-Mus81 antibody, Mouse monoclonal

clone MTA30 2G10/3, purified from hybridoma cell culture

Synonym(s):

Anti-SLX3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

MTA30 2G10/3, monoclonal

form

buffered aqueous solution

mol wt

antigen ~59 kDa

species reactivity

human, monkey

technique(s)

immunoprecipitation (IP): suitable
western blot: 0.5-1 μg/mL using HEK-293T total cell extract

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MUS81(80198)

General description

Monoclonal Anti-Mus81 (mouse IgG2a isotype) is derived from the hybridoma MTA30 2G10/3 produced by the fusion of mouse myeloma cells (Sp2/0 Ag.14 cells) and splenocytes from BALB/c mice immunized with Mus81 protein. Crossover junction endonuclease Mus81 protein was identified by screening a human cerebellum cDNA library with a probe from the yeast Mus81. The human Mus81 has 551 amino acids with a molecular mass of 59 kDa. The highest sequence similarity between different species is around the VERKX3D motif. This motif is conserved in the endonuclease family (XPF) family of nucleases. The protein has helix-hairpin-helix DNA-binding domains in the N- and C-termini of the protein that are also conserved.

Immunogen

Mus81 protein.

Application

Anti-Mus81 antibody, Mouse monoclonal has been used in immunofluorescence, immunoblotting and immunoprecipitation.

Biochem/physiol Actions

Crossover junction endonuclease Mus81 is elevated in cells exposed to agents that block DNA replication. Knockout mice that are homozygous Mus81-/-, or their heterozygous cohorts Mus81-/+, are susceptible to spontaneous chromosomal damage and cancer predisposition suggesting that this protein is important for genomic integrity and tumor suppression

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antonio Porro et al.
Nature communications, 8(1), 1073-1073 (2017-10-21)
Interstrand cross-link (ICL) hypersensitivity is a characteristic trait of Fanconi anemia (FA). Although FANCD2-associated nuclease 1 (FAN1) contributes to ICL repair, FAN1 mutations predispose to karyomegalic interstitial nephritis (KIN) and cancer rather than to FA. Thus, the biological role of
Regulation of multiple DNA repair pathways by the Fanconi anemia protein SLX4
Kim Y, et al.
Blood, 121(1), 54-63 (2013)
The Mus81 solution to resolution: generating meiotic crossovers without Holliday junctions
Hollingsworth NM and Brill SJ
Genes & Development, 18(2), 117-125 (2004)
Nadeem Shaikh et al.
Genome biology, 23(1), 223-223 (2022-10-22)
A major driver of cancer chromosomal instability is replication stress, the slowing or stalling of DNA replication. How replication stress and genomic instability are connected is not known. Aphidicolin-induced replication stress induces breakages at common fragile sites, but the exact
Oncogenes induce genotoxic stress by mitotic processing of unusual replication intermediates
Neelsen KJ, et al.
The Journal of Cell Biology, 200(6), 699-708 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service