Key Documents
M1435
Sigma-Aldrich
Microarray Spotting Solution for Silane Coated Slides
About This Item
UNSPSC Code:
12352200
Features and Benefits
Improves spot morphology and DNA binding to the slide resulting in increased hybridization signal.
Other Notes
Optimized spotting solution for use with SigmaScreen™ APS Coated Slides for Microarrays (Product Code S9936).
Choose from one of the most recent versions:
Certificates of Analysis (COA)