Skip to Content
Merck
All Photos(1)

Key Documents

M1014

Sigma-Aldrich

Methyl 3-amino-3-deoxy-α-D-mannopyranoside hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H15NO5 · HCl
CAS Number:
Molecular Weight:
229.66
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

form

solid

storage temp.

2-8°C

SMILES string

Cl.COC1OC(CO)C(O)C(N)C1O

InChI

1S/C7H15NO5.ClH/c1-12-7-6(11)4(8)5(10)3(2-9)13-7;/h3-7,9-11H,2,8H2,1H3;1H

InChI key

UMFRURBOSLCTIW-UHFFFAOYSA-N

Application

Methyl 3-amino-3-deoxy-α-D-mannopyranoside hydrochloride has been used in a study to assess the importance of the nitrogen protecting group on stereoselectivity. [1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David Crich et al.
The Journal of organic chemistry, 72(14), 5183-5192 (2007-06-15)
The highly stereocontrolled synthesis of the 3-amino-3-deoxy-beta-mannopyranosides is achieved by means of thioglycoside donors protected with a 4,6-O-benzylidene or alkylidene acetal and a benzylidene imine group. Among the various nitrogen protecting groups investigated only the Schiff's base was found to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service