M0671
Recombinant Molecular Weight Marker
mol wt 15,000-150,000 Da
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
usage
1 vial sufficient for 100 uses (10 × 10 cm mini gels)
1 vial sufficient for 50 uses (large format 16 × 18 cm gels)
Quality Level
mol wt
15,000-150,000 Da
storage temp.
2-8°C
Application
Recombinant Molecular Weight Marker has been used for determining the size of protein bands in western blot and sodium dodecyl sulfate polyacrylamide gel electrophoresis (SDS-PAGE).
Other Notes
The recombinant molecular weight standard for SDS-PAGE contains seven precisely sized proteins, M.W. 15, 25, 35, 50, 75, 100 and 150 kDa. Recombinant proteins produce sharp bands for precise size determination since they are not glycosylated.
Physical form
Solution containing approx. 0.8 mg/ml total protein in 125 mM Tris-HCl, pH 6.8, 2% SDS, 10% glycerol, 200 mM 2-mercaptoethanol, 0.007% bromphenol blue.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Sens. 1
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Cloning, expression and functional characterisation of chitinase from larvae of tomato moth (Lacanobia oleracea): a demonstration of the insecticidal activity of insect chitinase.
Insect Biochemistry and Molecular Biology, 34, 1037-1050 (2004)
Cell cycle dependent telomere regulation by telomerase in human bone marrow mesenchymal stem cells.
Biochemical and Biophysical Research Communications, 369, 1114-1119 (2008)
Clinical and vaccine immunology : CVI, 15(8), 1292-1300 (2008-06-27)
Earlier studies have demonstrated in A/Sn mice highly susceptible to Chagas' disease protective immunity against lethal Trypanosoma cruzi infection elicited by vaccination with an open reading frame (ORF) expressed by amastigotes. In our experiments, we used this mouse model to
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service