Skip to Content
Merck
All Photos(1)

Documents

LIBHV1

Sigma-Aldrich

MISSION® LentiElite Whole-Genome shRNA Library

Synonym(s):

Arrayed Whole Genome shRNA library, Arrayed library, Arrayed shRNA, Arrayed shRNA library, Human Arrayed Library, Human Library, Lentiviral Library, MISSION shRNA, Sigma shRNA Library, Whole Genome Library

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105904
NACRES:
NA.51

product line

MISSION®

technique(s)

capture ELISA: 1 × 106 TU/mL using p24

shipped in

dry ice

storage temp.

−70°C

General description

We, along with the RNAi Consortium (TRC) provide the human MISSION shRNA Library, developed at the Broad Institute, a joint venture between MIT and Harvard. The Complete Human (and mouse) MISSION shRNA Library (TRC 1, 1.5, and 2.0) is exclusive to us and contains almost 130,000 human shRNA clones and more than 43,000 validated clones. 

It combines all of the highly trusted TRC1 content plus an additional 49,000 clones targeting 4,000 new human genes. The MISSION shRNA Library (also available in mouse) is the most comprehensive and thoroughly validated shRNA collection available.

When working with non-transfectable or non-dividing cells, lentiviral shRNA libraries will provide you with efficient, long-term knockdown using a robust and quality system. Screen over 20,000 human genes using the LentiElite shRNA library.

Features and Benefits

Vector: pLKOVector (U6-shRNA- hPGK-Puro)
Coverage: 20,000+ human genes, 125,000+ shRNA Clones

Components

20 μL aliquot per lentiviral shRNA clone, -400 x 384 well plates, 10^6 titer

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

RNAi Consortium (TRC): Collaborative effort among academic labs and biotech/pharma institutes advancing RNA interference research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service