Skip to Content
Merck
All Photos(1)

Key Documents

L9793

Sigma-Aldrich

LP44

≥98% (HPLC), solid

Synonym(s):

4-[2-(Methylthio)phenyl]-N-(1,2,3,4-tetrahydro-1-naphth alenyl)-1-piperazinehexanamide hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$247.00
50 MG
$978.00

$247.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$247.00
50 MG
$978.00

About This Item

Empirical Formula (Hill Notation):
C27H38ClN3OS
CAS Number:
Molecular Weight:
488.13
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$247.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

solid

solubility

DMSO: >10 mg/mL
H2O: ≥2 mg/mL

storage temp.

2-8°C

SMILES string

Cl.CSc1ccccc1N2CCN(CCCCCC(=O)NC3CCCc4ccccc34)CC2

InChI

1S/C27H37N3OS.ClH/c1-32-26-15-7-6-14-25(26)30-20-18-29(19-21-30)17-8-2-3-16-27(31)28-24-13-9-11-22-10-4-5-12-23(22)24;/h4-7,10,12,14-15,24H,2-3,8-9,11,13,16-21H2,1H3,(H,28,31);1H

InChI key

DWGKCWWWKHCVDH-UHFFFAOYSA-N

Application

LP44 has been used as a 5-hydroxytryptamine (serotonin) receptor 7 (5-HT7) agonist to study its effects on mitochondria-mediated leucine-rich repeat and pyrin domain containing 3 (NLRP3) inflammasome activation.[1]

Biochem/physiol Actions

LP44 is a high affinity 5-HT7 receptor agonist.
LP44 is a high affinity 5-HT7 receptor agonist. Ki = 0.22 nM. LP44 displays selectivity over 5-HT1A and 5-HT2A receptors (200- and >1000-fold, respectively). LP44 induces relaxation of substance P-stimulated guinea pig ileum (EC50 = 2.56 μM).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miriam A Snider et al.
Journal of animal science, 96(4), 1570-1578 (2018-02-23)
Prior work using a contractility bioassay determined that the serotonin (5-HT) receptor subtype 5-HT2A is present in bovine lateral saphenous veins and plays a role in ergot alkaloid (EA)-induced vascular contraction in steers grazing endophyte-infected (Epichloë coenophiala) tall fescue (Lolium
Jianshu Ni et al.
BJU international, 123(4), 718-725 (2018-05-22)
To examine the effect of intrathecal (i.t.) serotonin (5-hydroxytryptamine) 5-HT7 agonist administration on voiding function in the urethane-anesthetised rat, and the change in 5-HT7 receptor (5-HT7 R) expression in the lumbosacral cord Onuf's nucleus after spinal cord injury (SCI). In
Lynn Meurs et al.
PloS one, 6(9), e24393-e24393 (2011-09-21)
Schistosoma infection is thought to lead to down-regulation of the host's immune response. This has been shown for adaptive immune responses, but the effect on innate immunity, that initiates and shapes the adaptive response, has not been extensively studied. In
Gulsen Ayaz et al.
Biochemical genetics, 55(1), 34-47 (2016-09-03)
This study aimed to investigate the effects of the 5-HT7 receptor agonist (LP44) and antagonist (SB269970) on LPS-induced in vivo tissue damage and cell culture by molecular methods. This study was conducted in two steps. For in vivo studies, 24 female
Liping Wu et al.
Journal of gastroenterology, 51(11), 1040-1049 (2016-10-28)
Serotonin regulates gastrointestinal function, and mast cells are a potential nonneuronal source of serotonin in the esophagus. Tight junction (TJ) proteins in the esophageal epithelium contribute to the barrier function, and the serotonin signaling pathway may contribute to epithelial leakage

Articles

We offer many products related to serotonin receptors for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service