Skip to Content
Merck
All Photos(1)

Key Documents

L6715

Sigma-Aldrich

Biocoll

liquid, sterile-filtered, BioReagent, suitable for hybridoma

Synonym(s):

Separation Medium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75
Pricing and availability is not currently available.

sterility

sterile-filtered
sterile

Quality Level

product line

BioReagent

form

liquid

technique(s)

cell culture | hybridoma: suitable

impurities

endotoxin, tested

density

1.077 g/mL at 25 °C

shipped in

ambient

storage temp.

room temp

Application

Used to create a density medium for the purification of lymphocytes and other mononuclear cells. Histopaque®-1077 is a solution of polysucrose and sodium diatrizoate, adjusted to a density of 1.077 +/- 0.001 g/ml. This medium facilitates rapid recovery of viable lymphocytes from small volumes of whole blood. This technique is suitable for use in cell mediated lympholysis and for human lymphocyte antigen (HLA)2 typing. It may also be employed as the initial isolation step prior to enumeration of T-, B-, and ‘null′ lymphocytes.

Legal Information

Histopaque is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service