Skip to Content
Merck
All Photos(1)

Key Documents

L5876

Sigma-Aldrich

D-Lysine monohydrochloride

≥98% (TLC)

Synonym(s):

(R)-2,6-Diaminohexanoic acid monohydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2N(CH2)4CH(NH2)CO2H·HCl
CAS Number:
Molecular Weight:
182.65
Beilstein:
4356907
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (TLC)

form

powder

color

white to off-white

mp

266 °C (dec.) (lit.)

solubility

H2O: soluble

application(s)

peptide synthesis

SMILES string

Cl[H].NCCCC[C@@H](N)C(O)=O

InChI

1S/C6H14N2O2.ClH/c7-4-2-1-3-5(8)6(9)10;/h5H,1-4,7-8H2,(H,9,10);1H/t5-;/m1./s1

InChI key

BVHLGVCQOALMSV-NUBCRITNSA-N

Looking for similar products? Visit Product Comparison Guide

Application

D-Lysine is used as a component of a wide array of polymers (poly-D-lysine), surfactants and biofilms to confer a positive (cationic) charge.

related product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sara Fornera et al.
Macromolecular bioscience, 11(8), 1052-1067 (2011-05-14)
Horseradish peroxidase (HRP) is immobilized in three easy steps on SiO(2) surfaces with the help of a polycationic second generation dendronized polymer (denpol) and the biotin-avidin system. This stepwise immobilization process is monitored and quantitatively analyzed with the transmission interferometric
Yanfei Yang et al.
Reproduction, fertility, and development, 23(3), 496-505 (2011-03-24)
Gonocytes are the only type of germ cells present in the postnatal testis and give rise to spermatogonial stem cells. Purification of gonocytes has important implications for the study and manipulation of these cells and may provide insights for the
Dhananjoy Mondal et al.
Carbohydrate research, 346(5), 588-594 (2011-02-15)
The synthesis of four dicationic glucose-templated D-lysine-derived surfactants and their two unmodified D-lysine-analogs is described. Replacement of D-lysine by D-glucose-templated-D-lysine provides a general tool to introduce chemical diversity into the side chain of lysine. The presence of the polyfunctional D-gluco-configured
Leslee Sprague et al.
BMC immunology, 12, 35-35 (2011-06-08)
Dendritic cells (DCs) are antigen presenting cells capable of inducing specific immune responses against microbial infections, transplant antigens, or tumors. Interestingly, microenvironment conditions such as those present in tumor settings might induce a DC phenotype that is poorly immunogenic and
Kyungtae Kang et al.
Biomaterials, 32(27), 6374-6380 (2011-06-10)
Chemical surface modification of neuron-surface interfaces is essential for the development of biologically active and functional neural interfaces. Different types of surface modification schemes are required to derivatize either electrode or insulator surfaces, which limits the surface chemistry based neural

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service