Skip to Content
Merck
All Photos(1)

Key Documents

L3040

Sigma-Aldrich

L-765,314 hydrate

≥98% (HPLC), solid

Synonym(s):

(2S)-4-(4-amino-6,7-dimethoxy-2-quinazolinyl)-2-[[(1,1-dimethylethyl)amino]carbonyl]-1-piperazinecarboxylic acid, phenylmethyl ester hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H34N6O5 · xH2O
Molecular Weight:
522.60 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (HPLC)

form

solid

solubility

DMSO: 14 mg/mL

originator

Merck & Co., Inc., Kenilworth, NJ, U.S.

SMILES string

O.COc1cc2nc(nc(N)c2cc1OC)N3CCN([C@@H](C3)C(=O)NC(C)(C)C)C(=O)OCc4ccccc4

InChI

1S/C27H34N6O5.H2O/c1-27(2,3)31-24(34)20-15-32(11-12-33(20)26(35)38-16-17-9-7-6-8-10-17)25-29-19-14-22(37-5)21(36-4)13-18(19)23(28)30-25;/h6-10,13-14,20H,11-12,15-16H2,1-5H3,(H,31,34)(H2,28,29,30);1H2/t20-;/m0./s1

InChI key

KQHMFDHSRXAELV-BDQAORGHSA-N

Biochem/physiol Actions

Potent and selective α1B-adrenoceptor antagonist.

Features and Benefits

This compound is featured on the α1-Adrenoceptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Merck & Co., Inc., Kenilworth, NJ, U.S.. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service