Skip to Content
Merck
All Photos(2)

Key Documents

L2885

Sigma-Aldrich

Lymphotoxin-β Receptor/Fc Chimera human

>95% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder, suitable for cell culture

Synonym(s):

TNF R-related protein, TNF RIII

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800

biological source

human

recombinant

expressed in NSO cells

Assay

>95% (SDS-PAGE)

form

lyophilized powder

mol wt

monomer calculated mol wt 49.6 kDa
70 kDa by SDS-PAGE (reducing) (due to glycosylation)

packaging

pkg of 100 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... LTBR(4055)

Biochem/physiol Actions

LTBR (lymphotoxin β receptor) is a receptor for LIGHT (tumor necrosis factor ligand superfamily member 14) and LT (lymphotoxin)-a1β2. It is present in the epithelial, stromal and myeloid cells. LTBR participates in lymphoid organ development and immune homeostasis. It is also involved in the lipopolysaccharide (LPS)-mediated expression of IL (interleukin)-8.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline.

Analysis Note

The biological activity is measured by the ability of the immobilized LTβR/Fc to bind LTα2/β1 and LTα1/β2 in a functional ELISA.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seok-Won Jang et al.
Immunology letters, 165(2), 63-69 (2015-04-19)
Lymphotoxin-beta receptor (LTβR), a receptor for LIGHT and LTα1β2, is expressed on the epithelial, stromal, and myeloid cells. LTβR is known to affect the lymphoid organ development and immune homeostasis. However, its role in macrophage function has not been sufficiently
M Baens et al.
Genomics, 16(1), 214-218 (1993-04-01)
An arrayed library of human heterogeneous nuclear complementary (hnc) DNA was constructed from a somatic cell hybrid (M28) containing an i(12p) marker as the sole human chromosome. Heterogeneous nuclear (hn) RNA of M28 was used to synthesize first-strand hncDNA with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service