Skip to Content

L1908

γ-O-Octadecyl-L-α-lysophosphatidylcholine

Synonym(s):

1-O-Octadecyl-sn-glycero-3-phosphocholine, Lyso-C18-platelet activating factor, Lyso-PAF-C18

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C26H56NO6P
CAS Number:
Molecular Weight:
509.70
UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


storage temp.

−20°C



Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library