The specific origin of the lectin is considered proprietary. However, this is not a recombinant source lectin; the material is extracted from lentils, a plant material.
L0511
Lectin from Lens culinaris (lentil)
Sepharose™ conjugate, saline suspension
Synonym(s):
Lens culinaris agglutinin, LcH
Select a Size
$743.00
Estimated to ship onJune 02, 2025
Select a Size
About This Item
$743.00
Estimated to ship onJune 02, 2025
Recommended Products
conjugate
Sepharose™ conjugate
Quality Level
form
saline suspension
extent of labeling
~2 mg per mL
matrix
Sepharose 4B
capacity
15 mg/mL binding capacity (porcine thyroglobulin)
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Related Categories
Biochem/physiol Actions
Physical form
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
104.0 °F - closed cup
Flash Point(C)
40 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Find the right lectin for your research with our lectin selection guide, organized by lectin source/species, carbohydrate specificity, blood group specificity, and more.
-
Hello, I just want to make sure that the lens culinaris agglutinin is animal component free. Best regards Bjørn
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service