Skip to Content
Merck
All Photos(1)

Key Documents

L0168

Sigma-Aldrich

LB Agar Ampicillin - 50, Plates

pre-poured LB agar plates with 50 μg/ml ampicillin

Synonym(s):

LB Agar Plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
sterility:
sterile; autoclaved
suitability:
selective for Escherichia coli
selective for coliforms
Pricing and availability is not currently available.

grade

Molecular Biology
for molecular biology

sterility

sterile; autoclaved

form

plates (10 cm)
plates (10cm, 20-25ml each)

availability

available only in USA

capacity

20-25 mL

storage temp.

2-8°C

suitability

selective for Escherichia coli
selective for coliforms

General description

Miller′s LB is a highly-referenced microbial growth medium used for the cultivation of E. coli. This nutrient-rich microbial broth contains peptides, amino acids, water-soluble vitamins, and carbohydrates in a low-salt formulation. The addition of agar provides a solid medium for microbial growth.

Application

Suitable for selective cultivation of E. coli strains that contain plasmids conferring ampicillin resistance. The bacteria can be used for DNA plasmid production or production of recombinant proteins.

Features and Benefits

Miller′s LB Agar plates provide:
  • Convenient, ready-to-use 10cm format
  • Standard formulation plus antibiotic

Components

15g/L Agar
10g/L Tryptone
10g/L NaCl
5g/L Yeast Extract
50μg/ml ampicillin

Other Notes

The plates must be used within two months of the manufacture date to ensure the antibiotics are fully active. The product may be shipped up until its expiration date but is generally shipped 2-4 weeks before expiration.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

Genetic engineering enables large-scale expression and isolation of recombinant proteins for research purposes.

Questions

1–2 of 2 Questions  
  1. Why do the LB-agar plates with antibiotics have such a short expiration date?

    1 answer
    1. The antibiotics in the LB-agar plates are not stable.  The antibiotic degrades over time, making it ineffective at selecting transformed bacteria grown on the plate.  Because of this, we recommend the plates are used as soon as possible once they are received.  For labs with low use, the plates may expire quickly, so they may benefit from pouring their own plates using the LB - Miller powder with agar formulation (L3147).

      Helpful?

  2. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service