Skip to Content
Merck
All Photos(1)

Key Documents

K1250

Sigma-Aldrich

6-Ketocholestanol

Synonym(s):

3β-Hydroxy-5α-cholestan-6-one, 5α-Cholestan-3β-ol-6-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H46O2
CAS Number:
Molecular Weight:
402.65
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

SMILES string

CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC(=O)[C@H]4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C

InChI

1S/C27H46O2/c1-17(2)7-6-8-18(3)21-9-10-22-20-16-25(29)24-15-19(28)11-13-27(24,5)23(20)12-14-26(21,22)4/h17-24,28H,6-16H2,1-5H3/t18-,19+,20+,21-,22+,23+,24-,26-,27-/m1/s1

InChI key

JQMQKOQOLPGBBE-ZNCJEFCDSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Valenta et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 57(2), 329-336 (2004-03-17)
The aim of this study was to investigate membrane interactions of phloretin and 6-ketocholestanol using different methods. A previously reported colorimetric assay with phospholipid/polydiacetylene (PDA) vesicles was used to examine a possible interaction of phloretin and 6-ketocholestanol with this target.
C Valenta et al.
International journal of pharmaceutics, 217(1-2), 79-86 (2001-04-09)
In this study the effect of phloretin (PH) and 6-ketocholestanol (KC) on the permeation of progesterone through porcine skin has been examined. Both PH and KC were incorporated into unilamellar L-alpha-phosphatidylcholine (PC) liposomes at different concentrations (7.5, 15, 30 and
Tudor Luchian et al.
Langmuir : the ACS journal of surfaces and colloids, 22(20), 8452-8457 (2006-09-20)
As a result of the interfacial chemical heterogenity, membrane-penetrating peptides will experience a dramatic variation in environmental polarity manifested via electrical interactions with the surface and dipole potential of membranes prone to modulate the membrane insertion and folding of different
J I Alakoskela et al.
Biophysical journal, 80(1), 294-304 (2001-02-13)
Nitro-2,1,3-benzoxadiazol-4-yl (NBD) group is a widely used, environment-sensitive fluorescent probe. The negatively charged dithionite rapidly reduces the accessible NBD-labeled lipids in liposomes to their corresponding nonfluorescent derivatives. In this study both the phospholipid headgroup and acyl chain NBD-labeled L-alpha-1,2-dipalmitoyl-sn-glycero-3-phospho-[N-(4-nitrobenz-2-oxa-1,3-diazole)-ethanolamine] (DPPN)
Xin-Min Li et al.
Biophysical journal, 85(6), 3788-3801 (2003-12-04)
Membrane microdomains, such as caveolae and rafts, are enriched in cholesterol and sphingomyelin, display liquid-ordered phase properties, and putatively function as protein organizing platforms. The goal of this investigation was to identify sterol and sphingomyelin structural features that modulate surface

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service