Skip to Content
Merck
All Photos(1)

Documents

J3770

Sigma-Aldrich

JNJ7777120

≥98% (HPLC)

Synonym(s):

1-[(5-Chloro-1H-indol-2-yl)carbonyl]-4-methyl-piperazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H16N3OCl
CAS Number:
Molecular Weight:
277.75
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

color

white to off-white

solubility

DMSO: >20 mg/mL
H2O: insoluble

originator

Johnson & Johnson

storage temp.

room temp

SMILES string

CN1CCN(CC1)C(=O)c2cc3cc(Cl)ccc3[nH]2

InChI

1S/C14H16ClN3O/c1-17-4-6-18(7-5-17)14(19)13-9-10-8-11(15)2-3-12(10)16-13/h2-3,8-9,16H,4-7H2,1H3

InChI key

HUQJRYMLJBBEDO-UHFFFAOYSA-N

Application

JNJ7777120 has been used as a histamine-4 receptor antagonist:
  • to study its effects on the pro-inflammatory microglia in rats
  • to study its effects on the Parkinson′s-like pathology in rat brain
  • to study its effects on the histamine receptor interaction in periodontal ligament fibroblasts (PDLF)

Biochem/physiol Actions

JNJ7777120 may exhibit neuroprotective effects against ischemic brain damage. It acts as an anti-inflammatory agent to treat inflammatory diseases. JNJ7777120 is observed to reduce colonic injury, cytokine production, and neutrophil infiltration.
JNJ7777120 is a potent, selective non-imidazole H4 histamine receptor antagonist.

Features and Benefits

This compound is featured on the Histamine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Johnson & Johnson. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Zampeli et al.
Inflammation research : official journal of the European Histamine Research Society ... [et al.], 58(6), 285-291 (2009-02-03)
Although the H(4) receptor localisation in the eye is unresolved, this study aimed to investigate the effects of the H(4) receptor antagonist JNJ7777120 in a model of experimental conjunctivitis. JNJ7777120, at 0.005-1 mmol/l, was instilled into the lower conjunctival fornix
Heiko Schenk et al.
Immunopharmacology and immunotoxicology, 38(5), 379-384 (2016-08-26)
The modulation of antigen uptake and activation of dendritic cells (DCs) by histamine may function as a regulator of inflammation. Therefore, we sought to determine the impact of histamine on antigen uptake by and activation of murine DCs. DCs from
Qiuyuan Fang et al.
Brain, behavior, and immunity, 92, 127-138 (2020-11-30)
Growing evidence indicates that microglia activation and a neuroinflammatory trigger contribute to dopaminergic cell loss in Parkinson's disease (PD). Furthermore, increased density of histaminergic fibers and enhanced histamine levels have been observed in the substantia nigra of PD-postmortem brains. Histamine-induced
Susanne Mommert et al.
International archives of allergy and immunology, 166(3), 225-230 (2015-05-01)
Natural killer (NK) cells have been detected in the lesional skin of patients with inflammatory skin diseases, where high levels of histamine are also present. Therefore, we investigated the effect of histamine, in particular via the histamine H4 receptor (H4R)
C Hoffmann et al.
Molecular pharmacology, 88(3), 552-560 (2015-07-15)
Over the past decade the kinetics of ligand binding to a receptor have received increasing interest. The concept of drug-target residence time is becoming an invaluable parameter for drug optimization. It holds great promise for drug development, and its optimization

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service