Skip to Content
Merck
All Photos(1)

Key Documents

J2753

Sigma-Aldrich

JWH-133

solid

Synonym(s):

3-(1,1-Dimethylbutyl)-1-deoxy-Δ8-THC, 3-(1,1-Dimethylbutyl)-1-deoxy-Δ8-tetrahydrocannabinol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H32O
CAS Number:
Molecular Weight:
312.49
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

drug control

USDEA Schedule I; Home Office Schedule 1; regulated under CDSA - not available from Sigma-Aldrich Canada

color

white

solubility

DMSO: <0.16 mg/mL
H2O: <0.28 mg/mL
ethanol: 11 mg/mL

storage temp.

−20°C

SMILES string

CCCC(C)(C)c1ccc2[C@@H]3CC(C)=CC[C@H]3C(C)(C)Oc2c1

Gene Information

Biochem/physiol Actions

Selective CB2 cannabinoid receptor agonist.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jörg Reifart et al.
Transplantation, 99(1), 41-47 (2014-11-02)
CD4+ T cells play a critical role during hepatic ischemia-reperfusion (I/R) injury although the mechanisms of their migration in the postischemic liver remain unclear. We answered the questions of whether hepatic stellate cells (HSCs) interact with CD4+ T cells during
Tianshui Yu et al.
Histology and histopathology, 30(6), 737-749 (2015-01-16)
The aim of the current study was to investigate the effects of cannabinoid receptor type 2 (CB2R) on the repair process of injured skeletal muscle, which could potentially lay solid foundations as a novel target for curing muscular fibrosis in
Weisi Fu et al.
Neuroscience letters, 595, 1-6 (2015-04-08)
Clinical trials investigating the analgesic efficacy of cannabinoids in multiple sclerosis have yielded mixed results, possibly due to psychotropic side effects mediated by cannabinoid CB1 receptors. We hypothesized that, a CB2-specific agonist (JWH-133) would decrease hyperalgesia in an experimental autoimmune
J W Huffman et al.
Bioorganic & medicinal chemistry, 7(12), 2905-2914 (2000-02-05)
The synthesis and pharmacology of 15 1-deoxy-delta8-THC analogues, several of which have high affinity for the CB2 receptor, are described. The deoxy cannabinoids include 1-deoxy-11-hydroxy-delta8-THC (5), 1-deoxy-delta8-THC (6), 1-deoxy-3-butyl-delta8-THC (7), 1-deoxy-3-hexyl-delta8-THC (8) and a series of 3-(1',1'-dimethylalkyl)-1-deoxy-delta8-THC analogues (2, n
Xiaoqing Li et al.
Toxicology letters, 313, 77-90 (2019-06-21)
Quetiapine is a common atypical antipsychotic used to treat mental disorders such as schizophrenia, bipolar disorder, and major depressive disorder. There has been increasing number of reports describing its cardiotoxicity. However, the molecular mechanisms underlying quetiapine-induced myocardial injury remain largely

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service