I9892
D-myo-Inositol 1,3,4-tris-phosphate hexapotassium salt
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C6H9O15P3K6
CAS Number:
Molecular Weight:
648.64
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
storage temp.
−20°C
SMILES string
[K+].[K+].[K+].[K+].[K+].[K+].O[C@@H]1[C@@H](O)[C@H](OP([O-])([O-])=O)[C@@H](OP([O-])([O-])=O)[C@@H](O)[C@H]1OP([O-])([O-])=O
InChI
1S/C6H15O15P3.6K/c7-1-2(8)5(20-23(13,14)15)6(21-24(16,17)18)3(9)4(1)19-22(10,11)12;;;;;;/h1-9H,(H2,10,11,12)(H2,13,14,15)(H2,16,17,18);;;;;;/q;6*+1/p-6/t1-,2-,3+,4+,5+,6+;;;;;;/m1....../s1
InChI key
DVOPWCOAGWTTEN-JQLBQNGOSA-H
Biochem/physiol Actions
Occurs naturally in carbachol-stimulated rat parotid glands1 and in adrenal glomerular cells.2
Quality
Contains excess sodium.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service