Skip to Content
Merck
All Photos(1)

Key Documents

I9785

Sigma-Aldrich

Imidazolo-oxindole PKR inhibitor C16

≥98% (HPLC)

Synonym(s):

6,8-Dihydro-8-(1H-imidazol-5-ylmethylene)-7H-pyrrolo[2,3-g]benzothiazol-7-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$133.00
25 MG
$526.00

$133.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$133.00
25 MG
$526.00

About This Item

Empirical Formula (Hill Notation):
C13H8N4OS
CAS Number:
Molecular Weight:
268.29
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$133.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

Assay

≥98% (HPLC)

color

yellow to orange-brown

solubility

DMSO: 12 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

O=C1NC2=CC=C3C(SC=N3)=C2/C1=C/C4=CNC=N4

InChI

1S/C13H8N4OS/c18-13-8(3-7-4-14-5-15-7)11-9(17-13)1-2-10-12(11)19-6-16-10/h1-6H,(H,14,15)(H,17,18)/b8-3-

InChI key

VFBGXTUGODTSPK-BAQGIRSFSA-N

Application

Imidazolo-oxindole PKR inhibitor C16 has been used:
  • to rescue fear memory deficits and restore long-term potentiation (LTP) impairment in mice[1]
  • to inhibit the strong induction of interferon stimulated genes (ISGs) after plasmid transfection in HeLa-S3 cells[2]
  • to augment eukaryotic translation initiation factor 2 (eIF2) activity[3]

Biochem/physiol Actions

C16 exhibits a neuroprotective role in adult brain injuries.[4]
Imidazolo-oxindole PKR inhibitor C16 is a selectiv inhibitor of RNA-dependent protein kinases (PKR, Eif2ak2).
Imidazolo-oxindole PKR inhibitor C16 is a selective inhibitor of RNA-dependent protein kinases (PKR, Eif2ak2). C16 is a first reported, potent and selective PKR inhibitor. Its inhibition effect on PKR is ATP-binding site directed. C16 specifically inhibits the apoptotic PKR/eIF2a signaling pathway without stimulating the proliferative mTOR/p70S6K signaling mechanism.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The specific protein kinase R (PKR) inhibitor C16 protects neonatal hypoxia-ischemia brain damages by inhibiting neuroinflammation in a neonatal rat model
Xiao J, et al.
Medical Science Monitor : International Medical Journal of Experimental and Clinical Research, 22, 5074-5074 (2016)
PKR-a Kinase to Remember
Gal-Ben-Ari S, et al.
Frontiers in Molecular Neuroscience, 11, 480-480 (2018)
AIM2 inflammasome mediates Arsenic-induced secretion of IL-1 beta and IL-18
Zhang MF, et al.
Oncoimmunology, 5(6), e1160182-e1160182 (2016)
Han Zhong et al.
Microbiology spectrum, 10(6), e0328222-e0328222 (2022-11-16)
African swine fever (ASF) is a highly contagious and often lethal disease of pigs caused by ASF virus (ASFV) and recognized as the biggest killer in global swine industry. Despite exhibiting incredible self-sufficiency, ASFV remains unconditionally dependent on the host
Mohamad A Zaini et al.
Scientific reports, 7, 42603-42603 (2017-02-16)
An important part of the beneficial effects of calorie restriction (CR) on healthspan and lifespan is mediated through regulation of protein synthesis that is under control of the mechanistic target of rapamycin complex 1 (mTORC1). As one of its activities

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service