Skip to Content
Merck
All Photos(1)

Key Documents

I9157

Sigma-Aldrich

Iodoplatinate spray reagent

Synonym(s):

Iodoplatinate Detection Spray

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$120.00

$120.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$120.00

About This Item

UNSPSC Code:
12352402
NACRES:
NA.26

$120.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

description

0.15% potassium chloroplatinate and 3% potassium iodide in dilute hydrochloric acid

technique(s)

thin layer chromatography (TLC): suitable

storage temp.

2-8°C

General description

Iodoplatinate spray reagent is suitable for the detection of alkaloids,[1] sulfur[2] amines and organic nitrogen containing compounds on Thin Layer Chromatography (TLC) plates.

Application

Iodoplatinate spray reagent has been used in the determination of radiochemical identity of N-13 Ammonia using Thin Layer Chromatography (TLC).[3]

Pictograms

Health hazardCorrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Met. Corr. 1 - STOT RE 2 Oral

Target Organs

Thyroid

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The
Peter JHS
Radiochemical Syntheses: Further Radiopharmaceuticals for Positron Emission Tomography and New Strategies for Their Production, 6, 671-676 (2015)
Bioautography guided screening of selected Indian medicinal plants reveals potent antimycobacterial activity of Allium sativum extracts-implication of non-sulfur compounds in inhibition.
Rajiniraja M and Jayaraman G
Leukemia, 6, 671-676 (2014)
Extraction of plant secondary metabolites
Natural Products, Volume 2 (2006)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service