Skip to Content
Merck
All Photos(1)

Key Documents

I7159

Sigma-Aldrich

Monoclonal Anti-Interleukin-4 antibody produced in mouse

clone 3010, purified immunoglobulin, lyophilized powder

Synonym(s):

Anti-IL-4

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41
conjugate:
unconjugated
application:
ELISA (c)
WB
clone:
3010, monoclonal
species reactivity:
human
citations:
7
technique(s):
capture ELISA: suitable
western blot: 1-2 μg/mL

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3010, monoclonal

form

lyophilized powder

species reactivity

human

technique(s)

capture ELISA: suitable
western blot: 1-2 μg/mL

isotype

IgG1

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... IL4(3565)

Looking for similar products? Visit Product Comparison Guide

General description

Interleukin-4 (IL-4) is a T cell derived chemokine that stimulates the Th2 mediated immune response and proliferation of B cells. The effects of IL-4 are mediated by two types of receptors, type I receptor consisting of IL-4Rα and common γ chain and type II receptor composed of IL-4Rα and IL-13Rα1. The signalling stimulated by IL-4 leads to activation of JAK/STAT6 and IRS-mediated PI3K/Akt pathway. Through these pathways, IL-4 is responsible for endocytic activity of macrophages, chemotaxis of leukocytes in response to inflammation, angiogenesis and regulation of nitric oxide metabolism in macrophages. Anti-tumor effects of IL-4 have been reported in cancers of breast, liver and renal cells
Anti-Interleukin-4 specifically recognizes human interleukin-4.

Immunogen

purified, E. coli-derived recombinant human interleukin-4.

Application

Anti-Interleukin-4 antibody may be used for immunoblotting at a working concentration of 1-2 μg/ml. For ELISA, a working concentration of 4 μg/ml is recommended.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Stein et al.
The Journal of experimental medicine, 176(1), 287-292 (1992-07-01)
Expression of the macrophage mannose receptor is inhibited by interferon gamma (IFN-gamma), a T helper type 1 (Th-1)-derived lymphokine. Interleukin 4 (IL-4), a Th-2 lymphocyte product, upregulates major histocompatibility class II antigen expression but inhibits inflammatory cytokine production by macrophages.
C K Oh et al.
European respiratory review : an official journal of the European Respiratory Society, 19(115), 46-54 (2010-10-20)
Asthma is a complex, persistent, inflammatory disease characterised by airway hyperresponsiveness in association with airway inflammation. Studies suggest that regular use of high-dose inhaled corticosteroids and long-acting bronchodilators or omalizumab (a humanised monoclonal antibody that binds to immunoglobulin E and
Jane Gilmour et al.
Immunology, 124(4), 437-444 (2008-05-16)
The mechanism of differentiation of naïve T cells to a variety of effector lineages, but particularly to T helper type 1 (Th1) and Th2 cells, has been the subject of intense scrutiny over the past two decades. Studies have revealed
Hao-Wei Wang et al.
Cell cycle (Georgetown, Tex.), 9(24), 4824-4835 (2010-12-15)
Although macrophages were originally recognized as major immune effector cells, it is now appreciated that they also play many important roles in the maintenance of tissue homeostasis, and are involved in a variety of pathological conditions including cancer. Several studies
H Jiang et al.
The Journal of allergy and clinical immunology, 105(6 Pt 1), 1063-1070 (2000-06-16)
In the past several years, extensive studies on the mechanisms underlying IL-4 and IL-13 signaling have enabled us to gain insight into how these cytokines regulate immune responses. Because both IL-4 and IL-13 use the IL-4Ralpha as a receptor component

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service