Skip to Content
Merck
All Photos(1)

Key Documents

I6410

Sigma-Aldrich

ISCK03

≥98% (HPLC), solid

Synonym(s):

Benzenesulfonamide,4-(1,1-dimethylethyl)-N-[4-(1H-imidazol-1-yl)phenyl], [4-t-Butylphenyl]-N-(4-imidazol-1-yl phenyl)sulfonamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$153.00
25 MG
$470.00

$153.00


Estimated to ship onMay 06, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$153.00
25 MG
$470.00

About This Item

Empirical Formula (Hill Notation):
C19H21N3O2S
CAS Number:
Molecular Weight:
355.45
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$153.00


Estimated to ship onMay 06, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

solid

color

yellow

solubility

DMSO: ≥20 mg/mL
H2O: insoluble

storage temp.

2-8°C

SMILES string

CC(C)(C)c1ccc(cc1)S(=O)(=O)Nc2ccc(cc2)-n3ccnc3

InChI

1S/C19H21N3O2S/c1-19(2,3)15-4-10-18(11-5-15)25(23,24)21-16-6-8-17(9-7-16)22-13-12-20-14-22/h4-14,21H,1-3H3

InChI key

XQABBHBFHWHMKF-UHFFFAOYSA-N

Biochem/physiol Actions

ISCK03 inhibits in vitro phosphorylation of c-Kit in a dose dependent manner.
ISCK03 inhibits in vitro phosphorylation of c-Kit in a dose dependent manner. ISCK03 inhibits SCF (stem cell factor)-induced c-kit phosphorylation in 501mel human melanoma cells. Complete inhibition was observed with 1 and 5 μM ISCK03.

Features and Benefits

This compound is featured on the PDGFR page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiroko Morimoto et al.
PloS one, 4(11), e7909-e7909 (2009-11-26)
Spermatogonial stem cells (SSCs) continuously undergo self-renewal division to support spermatogenesis. SSCs are thought to have a fixed phenotype, and development of a germ cell transplantation technique facilitated their characterization and prospective isolation in a deterministic manner; however, our in
Yuan Ma et al.
Cell cycle (Georgetown, Tex.), 19(16), 2074-2080 (2020-07-14)
Neurogenic bladder (NB) is a type of double renal dysfunction caused by nerve lesions. The interstitial cells of Cajal (ICC) damage are involved in bladder dysfunction. The aim of this study is to investigate the effect of stem cell factor
Jin Man Kim et al.
Development (Cambridge, England), 148(6) (2021-03-05)
The actomyosin complex plays crucial roles in various life processes by balancing the forces generated by cellular components. In addition to its physical function, the actomyosin complex participates in mechanotransduction. However, the exact role of actomyosin contractility in force transmission
Genevra Kuziel et al.
Cancers, 12(8) (2020-08-01)
Obesity is correlated with breast tumor desmoplasia, leading to diminished chemotherapy response and disease-free survival. Obesity causes chronic, macrophage-driven inflammation within breast tissue, initiated by chemokine ligand 2 (CCL2) signaling from adipose stromal cells. To understand how CCL2-induced inflammation alters
Cheong-Yong Yun et al.
Theranostics, 10(1), 340-352 (2020-01-07)
Rationale: Microphthalmia-associated transcription factor M (MITF-M) plays important roles in the pigment production, differentiation and survival of melanocytes. Stem cell factor (SCF) and its receptor KIT stimulate MITF-M activity via phosphorylation at the post-translation level. However, the phosphorylation shortens half-life

Articles

We offers many products related to PDGFR for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service