Skip to Content
Merck
All Photos(1)

Key Documents

I5404

Sigma-Aldrich

IgG2, Kappa from human myeloma plasma

purified immunoglobulin, >95% (SDS-PAGE)

Synonym(s):

Human IgG2-κ

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$270.00

$270.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$270.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

$270.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

biological source

human

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

Assay

>95% (SDS-PAGE)

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

IgG antibody subtype is the most abundant serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defence of the neonate against infections.[1]
Human myeloma IgG2, κ is purified from human plasma by fractionation, ion-exchange and affinity chromatography procedures.

Application

The purified IgG2, κ may be used as an immunoglobulin calibrator, reference antigen, blocking agent or coating protein in a variety of immunoassays including ELISA, dot-blot immunobinding, Western immunoblotting, immunodiffusion, immunoelectrophoresis, hemagglutination, and cell-binding assays. Human myeloma IgG2, κa was used to characterize the trisulfide modifications in human and mouse antibodies[2] and in flow cytometry.[3]

Physical form

Solution in 0.02 M Tris buffered saline, pH 8.0.

Analysis Note

Identity is determined by immunoelectrophoresis and indirect ELISA.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jorn J Heeringa et al.
Allergy, 75(5), 1121-1132 (2019-10-07)
While treatment for atopic rhinitis is aimed mostly to relieve symptoms, only allergen-specific immunotherapy (AIT) is targeted to modify the natural history of allergic diseases. This results in sustained clinical tolerance, even when treatment has stopped. The immunomodulatory effects of
Jonatan Dewulf et al.
Cancers, 13(9) (2021-05-06)
The involvement of RANK/RANKL signaling in the tumor microenvironment (TME) in driving response or resistance to immunotherapy has only very recently been recognized. Current quantification methods of RANKL expression suffer from issues such as sensitivity, variability, and uncertainty on the
Sheng Gu et al.
Analytical biochemistry, 400(1), 89-98 (2010-01-21)
Trisulfides are a posttranslational modification formed by the insertion of a sulfur atom into a disulfide bond. Although reports for trisulfides in proteins are limited, we find that they are a common modification in natural and recombinant antibodies of all
Misty W Stevens et al.
mAbs, 6(2), 547-555 (2014-02-05)
Ch-mAb7F9, a human-mouse chimeric monoclonal antibody (mAb) designed to bind (+)-methamphetamine (METH) with high affinity and specificity, was produced as a treatment medication for METH abuse. In these studies, we present the preclinical characterization that provided predictive evidence that ch-mAb7F9
Zhongqi Zhang et al.
mAbs, 11(8), 1381-1390 (2019-08-15)
Human IgG antibodies containing terminal alpha 2,6-linked sialic acid on their Fc N-glycans have been shown to reduce antibody-dependent cell-mediated cytotoxicity and possess anti-inflammatory properties. Although terminal sialylation on complex N-glycans can happen via either an alpha 2,3-linkage or an

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service