Skip to Content
Merck
All Photos(1)

Key Documents

I4879

Sigma-Aldrich

L-Isoleucine β-naphthylamide

≥98% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H20N2O
CAS Number:
Molecular Weight:
256.34
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26
Pricing and availability is not currently available.

Product Name

L-Isoleucine β-naphthylamide,

Assay

≥98% (TLC)

Quality Level

form

powder

color

white to off-white

storage temp.

2-8°C

SMILES string

CC[C@H](C)[C@H](N)C(=O)Nc1ccc2ccccc2c1

InChI

1S/C16H20N2O/c1-3-11(2)15(17)16(19)18-14-9-8-12-6-4-5-7-13(12)10-14/h4-11,15H,3,17H2,1-2H3,(H,18,19)/t11-,15-/m0/s1

InChI key

CEZPKIGJZWWHJT-NHYWBVRUSA-N

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service